accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHNNNHNOCH3

CAS RN 74150-27-9

IUPAC Name: 6-[2-(4-methoxyphenyl)-1H-1,3-benzodiazol-6-yl]-5-methyl-2,3,4,5-tetrahydropyridazin-3-one
Synonyms: pimobendan
Molecular Weight (g/mol): 334.38
Molecular Formula: C19H18N4O2
InChi Key: GLBJJMFZWDBELO-UHFFFAOYNA-N
SMILES: COC1=CC=C(C=C1)C1=NC2=CC=C(C=C2N1)C1=NNC(=O)CC1C
Grade 
  • (4)
Percent Purity 
  • (4)
Physical Form 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (4)
  • (2)
  • (4)
  • (4)
  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)