accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7418-65-7

NH2NHOO

CAS RN 7418-65-7

IUPAC Name: 4-aminopyridine-3-carboxylic acid
Synonyms: 4-aminopyridine-3-carboxylic acid
Molecular Weight (g/mol): 138.13
Molecular Formula: C6H6N2O2
InChi Key: IASBMUIXBJNMDW-UHFFFAOYSA-N
SMILES: NC1=CC=NC=C1C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results