accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOSOOHNOHNNCH3NOH3CN

CAS RN 74223-64-6

IUPAC Name: methyl 2-({[(4-methoxy-6-methyl-1,3,5-triazin-2-yl)carbamoyl]amino}sulfonyl)benzoate
Synonyms: metsulfuron-methyl
Molecular Weight (g/mol): 381.36
Molecular Formula: C14H15N5O6S
InChi Key: RSMUVYRMZCOLBH-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CC=C1S(=O)(=O)NC(=O)NC1=NC(C)=NC(OC)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results