accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7423-32-7

IUPAC Name: disodium dodecyl phosphate
Synonyms: disodium lauryl phosphate
Molecular Weight (g/mol): 310.28
Molecular Formula: C12H25Na2O4P
InChi Key: YVIGPQSYEAOLAD-UHFFFAOYSA-L
SMILES: [Na+].[Na+].CCCCCCCCCCCCOP([O-])([O-])=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results