accessibility menu, dialog, popup

UserName

Viewing profile as user:
(?)

CAS RN 7424-70-6

IUPAC Name: 9-(naphthalen-1-yl)anthracene
Synonyms: 9-(naphthalen-1-yl)anthracene
Molecular Weight (g/mol): 304.39
Molecular Formula: C24H16
InChi Key: YPNZWHZIYLWEDR-UHFFFAOYSA-N
SMILES: C1=CC2=CC3=CC=CC=C3C(C3=CC=CC4=CC=CC=C34)=C2C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results