CAS RN 7424-72-8
IUPAC Name:
9-(naphthalen-2-yl)anthracene
Synonyms:
9-(naphthalen-2-yl)anthracene
Molecular Weight (g/mol):
304.39
Molecular Formula:
C24H16
InChi Key:
MFDORGWIGJJZEQ-UHFFFAOYSA-N
SMILES:
C1=CC2=CC3=CC=CC=C3C(C3=CC4=CC=CC=C4C=C3)=C2C=C1
1
–
1
of
1
results
1
9-(2-Naphthyl)anthracene 98.0+%, TCI America™
CAS: 7424-72-8 Molecular Formula: C24H16 Molecular Weight (g/mol): 304.392 MDL Number: MFCD18205835 InChI Key: MFDORGWIGJJZEQ-UHFFFAOYSA-N PubChem CID: 264416 IUPAC Name: 9-naphthalen-2-ylanthracene SMILES: C1=CC=C2C=C(C=CC2=C1)C3=C4C=CC=CC4=CC5=CC=CC=C53