accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 74266-66-3

IUPAC Name: 5-bromo-1-fluoro-2-methoxy-3-nitrobenzene
Synonyms: 5-bromo-1-fluoro-2-methoxy-3-nitrobenzene
Molecular Weight (g/mol): 250.02
Molecular Formula: C7H5BrFNO3
InChi Key: CVCAYLYLJKBSNV-UHFFFAOYSA-N
SMILES: COC1=C(F)C=C(Br)C=C1[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results