accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2H2NOHOHSOOHO

CAS RN 74283-34-4

IUPAC Name: 2,4-diaminophenol; sulfuric acid
Synonyms: amidol; sulfuric acid
Molecular Weight (g/mol): 222.22
Molecular Formula: C6H10N2O5S
InChi Key: JKMWKYDJCPSJSI-UHFFFAOYSA-N
SMILES: OS(O)(=O)=O.NC1=CC(N)=C(O)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results