CAS RN 7429-44-9
IUPAC Name:
2-methoxycyclohexan-1-one
Synonyms:
2-methoxycyclohexan-1-one
Molecular Weight (g/mol):
128.17
Molecular Formula:
C7H12O2
InChi Key:
JYJURPHZXCLFDX-UHFFFAOYNA-N
SMILES:
COC1CCCCC1=O
1
–
1
of
1
results
1
2-Methoxycyclohexanone 95.0+%, TCI America™
CAS: 7429-44-9 Molecular Formula: C7H12O2 Molecular Weight (g/mol): 128.171 MDL Number: MFCD00009610 InChI Key: JYJURPHZXCLFDX-UHFFFAOYSA-N PubChem CID: 110998 IUPAC Name: 2-methoxycyclohexan-1-one SMILES: COC1CCCCC1=O