accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7429-91-6

IUPAC Name: dysprosium
Synonyms: dysprosium|disprosio|dysprosium atom
Molecular Weight (g/mol): 162.5
Molecular Formula: Dy
InChi Key: KBQHZAAAGSGFKK-UHFFFAOYSA-N
SMILES: [Dy]
Molecular Weight (g/mol) 
  • (28)
  • (29)
Quantity 
  • (24)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (3)
  • (2)
  • (4)
  • (1)
  • (2)
  • (4)
Percent Purity 
  • (5)
  • (1)
  • (14)
  • (4)
  • (18)
  • (2)
Form 
  • (4)
  • (3)
  • (2)
  • (5)
  • (4)
Physical Form 
  • (4)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (28)
  • (2)
  • (27)
  • (28)
  • (29)
  • (24)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (3)
  • (2)
  • (4)
  • (1)
  • (2)
  • (4)
  • (5)
  • (1)
  • (14)
  • (4)
  • (18)
  • (2)
  • (4)
  • (3)
  • (2)
  • (5)
  • (4)
  • (4)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)