accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 74370-93-7

IUPAC Name: 4-tert-butyl-1,3-thiazol-2-amine
Synonyms: 4-tert-butyl-1,3-thiazol-2-amine
Molecular Weight (g/mol): 156.25
Molecular Formula: C7H12N2S
InChi Key: CUWZBHVYLVGOAB-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CSC(N)=N1
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (6)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)