CAS RN 7439-89-6
Iron AA Standard, 1000 ppm Fe in 3% HNO3, Ricca Chemical
CAS: 7439-89-6 Molecular Formula: Fe Molecular Weight (g/mol): 55.85 MDL Number: MFCD00010999 InChI Key: XEEYBQQBJWHFJM-UHFFFAOYSA-N PubChem CID: 23925 ChEBI: CHEBI:82664 IUPAC Name: iron SMILES: [Fe]
Iron powder, -200 mesh, 99+% (metals basis), Thermo Scientific Chemicals
CAS: 7439-89-6 Molecular Formula: Fe Molecular Weight (g/mol): 55.85 MDL Number: MFCD00010999 InChI Key: XEEYBQQBJWHFJM-UHFFFAOYSA-N Synonym: ferrum,iron, elemental,ferrous,remko,armco,ferrovac,hoeganaes eh,3zhp,ancor b,atomiron 5m PubChem CID: 23925 ChEBI: CHEBI:82664 IUPAC Name: iron SMILES: [Fe]
Iron powder, spherical, <20 micron, 99.5% (metals basis), Thermo Scientific Chemicals
CAS: 7439-89-6 Molecular Formula: Fe Molecular Weight (g/mol): 55.85 MDL Number: MFCD00010999 InChI Key: XEEYBQQBJWHFJM-UHFFFAOYSA-N Synonym: ferrum,iron, elemental,ferrous,remko,armco,ferrovac,hoeganaes eh,3zhp,ancor b,atomiron 5m PubChem CID: 23925 ChEBI: CHEBI:82664 IUPAC Name: iron SMILES: [Fe]
Iron powder, spherical, 1-3 micron, 98+%, Thermo Scientific Chemicals
CAS: 7439-89-6 Molecular Formula: Fe Molecular Weight (g/mol): 55.85 MDL Number: MFCD00010999 InChI Key: XEEYBQQBJWHFJM-UHFFFAOYSA-N Synonym: ferrum,iron, elemental,ferrous,remko,armco,ferrovac,hoeganaes eh,3zhp,ancor b,atomiron 5m PubChem CID: 23925 ChEBI: CHEBI:82664 IUPAC Name: iron SMILES: [Fe]
Iron granules, 1-2mm (0.04-0.08in), 99.98% (metals basis)
CAS: 7439-89-6 Molecular Formula: Fe Molecular Weight (g/mol): 55.85 MDL Number: MFCD00010999 InChI Key: XEEYBQQBJWHFJM-UHFFFAOYSA-N Synonym: ferrum,iron, elemental,ferrous,remko,armco,ferrovac,hoeganaes eh,3zhp,ancor b,atomiron 5m PubChem CID: 23925 ChEBI: CHEBI:82664 IUPAC Name: iron SMILES: [Fe]
Iron foil, 0.1mm (0.004in) thick, 99.99% (metals basis)
CAS: 7439-89-6 Molecular Formula: Fe Molecular Weight (g/mol): 55.85 MDL Number: MFCD00010999 InChI Key: XEEYBQQBJWHFJM-UHFFFAOYSA-N Synonym: ferrum,iron, elemental,ferrous,remko,armco,ferrovac,hoeganaes eh,3zhp,ancor b,atomiron 5m PubChem CID: 23925 ChEBI: CHEBI:82664 IUPAC Name: iron SMILES: [Fe]
Iron AA Standard, Certified, 1000ppm ±5ppm (1mL = 1mg Fe), LabChem™
CAS: 7439-89-6 Molecular Formula: Fe Molecular Weight (g/mol): 55.85 MDL Number: MFCD00010999 InChI Key: XEEYBQQBJWHFJM-UHFFFAOYSA-N PubChem CID: 23925 ChEBI: CHEBI:82664 IUPAC Name: iron SMILES: [Fe]
Iron pieces, irregularly shaped, 25mm (1.0in) & down, 99.97+% (metals basis)
CAS: 7439-89-6 Molecular Formula: Fe Molecular Weight (g/mol): 55.85 MDL Number: MFCD00010999 InChI Key: XEEYBQQBJWHFJM-UHFFFAOYSA-N Synonym: ferrum,iron, elemental,ferrous,remko,armco,ferrovac,hoeganaes eh,3zhp,ancor b,atomiron 5m PubChem CID: 23925 ChEBI: CHEBI:82664 IUPAC Name: iron SMILES: [Fe]
Iron Metal, Reduced, 100 Mesh, Powder, Spectrum™ Chemical
CAS: 7439-89-6 Molecular Weight (g/mol): 55.85
Iron sputtering target, 50.8mm (2.0in) dia x 3.18mm (0.125in) thick, 99.95% (metals basis)
CAS: 7439-89-6 Molecular Formula: Fe Molecular Weight (g/mol): 55.85 MDL Number: MFCD00010999 InChI Key: XEEYBQQBJWHFJM-UHFFFAOYSA-N Synonym: ferrum,iron, elemental,ferrous,remko,armco,ferrovac,hoeganaes eh,3zhp,ancor b,atomiron 5m PubChem CID: 23925 ChEBI: CHEBI:82664 IUPAC Name: iron SMILES: [Fe]
Iron wire, 0.075mm (0.003in) dia, Puratronic™, 99.995% (metals basis)
CAS: 7439-89-6 Molecular Formula: Fe Molecular Weight (g/mol): 55.85 MDL Number: MFCD00010999 InChI Key: XEEYBQQBJWHFJM-UHFFFAOYSA-N Synonym: ferrum,iron, elemental,ferrous,remko,armco,ferrovac,hoeganaes eh,3zhp,ancor b,atomiron 5m PubChem CID: 23925 ChEBI: CHEBI:82664 IUPAC Name: iron SMILES: [Fe]
Iron foil, 0.5mm (0.02in) thick, 99.99% (metals basis)
CAS: 7439-89-6 Molecular Formula: Fe Molecular Weight (g/mol): 55.85 MDL Number: MFCD00010999 InChI Key: XEEYBQQBJWHFJM-UHFFFAOYSA-N Synonym: ferrum,iron, elemental,ferrous,remko,armco,ferrovac,hoeganaes eh,3zhp,ancor b,atomiron 5m PubChem CID: 23925 ChEBI: CHEBI:82664 IUPAC Name: iron SMILES: [Fe]
Iron slug, 6.35mm (0.25 in.) dia. x 12.7mm (0.50 in.) length, 99.95% (metals basis), Thermo Scientific Chemicals
CAS: 7439-89-6 Molecular Formula: Fe Molecular Weight (g/mol): 55.85 MDL Number: MFCD00010999 InChI Key: XEEYBQQBJWHFJM-UHFFFAOYSA-N Synonym: ferrum,iron, elemental,ferrous,remko,armco,ferrovac,hoeganaes eh,3zhp,ancor b,atomiron 5m PubChem CID: 23925 ChEBI: CHEBI:82664 IUPAC Name: iron SMILES: [Fe]
Iron slug, 3.175mm (0.125in) dia x 3.175mm (0.125in) length, 99.95% (metals basis)
CAS: 7439-89-6 Molecular Formula: Fe Molecular Weight (g/mol): 55.85 MDL Number: MFCD00010999 InChI Key: XEEYBQQBJWHFJM-UHFFFAOYSA-N Synonym: ferrum,iron, elemental,ferrous,remko,armco,ferrovac,hoeganaes eh,3zhp,ancor b,atomiron 5m PubChem CID: 23925 ChEBI: CHEBI:82664 IUPAC Name: iron SMILES: [Fe]
Iron wire, 0.25mm (0.01in) dia, Puratronic™, 99.99% (metals basis)
CAS: 7439-89-6 Molecular Formula: Fe Molecular Weight (g/mol): 55.85 MDL Number: MFCD00010999 InChI Key: XEEYBQQBJWHFJM-UHFFFAOYSA-N Synonym: ferrum,iron, elemental,ferrous,remko,armco,ferrovac,hoeganaes eh,3zhp,ancor b,atomiron 5m PubChem CID: 23925 ChEBI: CHEBI:82664 IUPAC Name: iron SMILES: [Fe]