CAS RN 7439-94-3
Lutetium pieces, 99.9% (REO)
CAS: 7439-94-3 Molecular Formula: Lu Molecular Weight (g/mol): 174.97 MDL Number: MFCD00011098 InChI Key: OHSVLFRHMCKCQY-UHFFFAOYSA-N Synonym: lutecium,unii-5h0doz21uj,5h0doz21uj,foil,foil, 0.127mm 0.005in thick reo,cassiopium,lutecio,cassiopeium,atom,pieces PubChem CID: 23929 ChEBI: CHEBI:33382 IUPAC Name: lutetium SMILES: [Lu]
Lutetium powder, -40 mesh, 99.9% (REO)
CAS: 7439-94-3 Molecular Formula: Lu Molecular Weight (g/mol): 174.97 MDL Number: MFCD00011098 InChI Key: OHSVLFRHMCKCQY-UHFFFAOYSA-N Synonym: lutecium,unii-5h0doz21uj,5h0doz21uj,foil,foil, 0.127mm 0.005in thick reo,cassiopium,lutecio,cassiopeium,atom,pieces PubChem CID: 23929 ChEBI: CHEBI:33382 IUPAC Name: lutetium SMILES: [Lu]
Lutetium pieces, distilled dendritic, 99.9% (REO)
CAS: 7439-94-3 Molecular Formula: Lu Molecular Weight (g/mol): 174.97 MDL Number: MFCD00011098 InChI Key: OHSVLFRHMCKCQY-UHFFFAOYSA-N Synonym: lutecium,unii-5h0doz21uj,5h0doz21uj,foil,foil, 0.127mm 0.005in thick reo,cassiopium,lutecio,cassiopeium,atom,pieces PubChem CID: 23929 ChEBI: CHEBI:33382 IUPAC Name: lutetium SMILES: [Lu]
Lutetium foil, 0.1mm (0.004in) thick, 99.9% (REO), Thermo Scientific Chemicals
CAS: 7439-94-3 Molecular Formula: Lu Molecular Weight (g/mol): 174.97 MDL Number: MFCD00011098 InChI Key: OHSVLFRHMCKCQY-UHFFFAOYSA-N Synonym: lutecium,unii-5h0doz21uj,5h0doz21uj,foil,foil, 0.127mm 0.005in thick reo,cassiopium,lutecio,cassiopeium,atom,pieces PubChem CID: 23929 ChEBI: CHEBI:33382 IUPAC Name: lutetium SMILES: [Lu]
Lutetium Standard For ICP, 1000μg/mL, SPEX CertiPrep™
CAS: 7439-94-3 Molecular Formula: Lu Molecular Weight (g/mol): 174.97 MDL Number: MFCD00011098 InChI Key: OHSVLFRHMCKCQY-UHFFFAOYSA-N PubChem CID: 23929 ChEBI: CHEBI:33382 IUPAC Name: lutetium SMILES: [Lu]
Lutetium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™
CAS: 7439-94-3 Molecular Formula: Lu Molecular Weight (g/mol): 174.97 MDL Number: MFCD00011098 InChI Key: OHSVLFRHMCKCQY-UHFFFAOYSA-N PubChem CID: 23929 ChEBI: CHEBI:33382 IUPAC Name: lutetium SMILES: [Lu]
Sigma Aldrich Lutetium
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Lutetium
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Lutetium
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Lutetium
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Lutetium
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Lutetium
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Lutetium
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Lutetium
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Lutetium
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
- 1
- 2