CAS RN 7439-96-5
Manganese standard solution, for AAS, 1 mg/ml Mn in 2 to 5% HNO3
CAS: 7439-96-5 | Mn | 54.94 g/mol
Manganese pieces, irregular, 99.9% (metals basis)
CAS: 7439-96-5 Molecular Formula: Mn Molecular Weight (g/mol): 54.94 MDL Number: MFCD00011111 InChI Key: PWHULOQIROXLJO-UHFFFAOYSA-N Synonym: colloidal,mangan,manganese, elemental,cutaval,magnacat,metal alloy,tronamang,fume,mangan polish,compounds PubChem CID: 23930 ChEBI: CHEBI:35154 IUPAC Name: manganese SMILES: [Mn]
Manganese, plasma standard solution, Specpure™ Mn 1000μg/mL
CAS: 7439-96-5 Molecular Formula: Mn Molecular Weight (g/mol): 54.94 MDL Number: MFCD00011111 InChI Key: PWHULOQIROXLJO-UHFFFAOYSA-N PubChem CID: 23930 ChEBI: CHEBI:35154 IUPAC Name: manganese SMILES: [Mn]
Manganese powder, -325 mesh, 99.3% (metals basis)
CAS: 7439-96-5 Molecular Formula: Mn Molecular Weight (g/mol): 54.94 MDL Number: MFCD00011111 InChI Key: PWHULOQIROXLJO-UHFFFAOYSA-N Synonym: colloidal,mangan,manganese, elemental,cutaval,magnacat,metal alloy,tronamang,fume,mangan polish,compounds PubChem CID: 23930 ChEBI: CHEBI:35154 IUPAC Name: manganese SMILES: [Mn]
Manganese pieces, 10mm & down, ultrapure, 99.9998% (metals basis)
CAS: 7439-96-5 Molecular Formula: Mn Molecular Weight (g/mol): 54.94 MDL Number: MFCD00011111 InChI Key: PWHULOQIROXLJO-UHFFFAOYSA-N Synonym: colloidal,mangan,manganese, elemental,cutaval,magnacat,metal alloy,tronamang,fume,mangan polish,compounds PubChem CID: 23930 ChEBI: CHEBI:35154 IUPAC Name: manganese SMILES: [Mn]
Manganese powder, -140+325 mesh, 99.6% (metals basis)
CAS: 7439-96-5 Molecular Formula: Mn Molecular Weight (g/mol): 54.94 MDL Number: MFCD00011111 InChI Key: PWHULOQIROXLJO-UHFFFAOYSA-N Synonym: colloidal,mangan,manganese, elemental,cutaval,magnacat,metal alloy,tronamang,fume,mangan polish,compounds PubChem CID: 23930 ChEBI: CHEBI:35154 IUPAC Name: manganese SMILES: [Mn]
Manganese pieces, irregular, 99.95% (metals basis)
CAS: 7439-96-5 Molecular Formula: Mn Molecular Weight (g/mol): 54.94 MDL Number: MFCD00011111 InChI Key: PWHULOQIROXLJO-UHFFFAOYSA-N Synonym: colloidal,mangan,manganese, elemental,cutaval,magnacat,metal alloy,tronamang,fume,mangan polish,compounds PubChem CID: 23930 ChEBI: CHEBI:35154 IUPAC Name: manganese SMILES: [Mn]
Manganese nanopowder, APS 30-50nm
CAS: 7439-96-5 Molecular Formula: Mn Molecular Weight (g/mol): 54.94 MDL Number: MFCD00011111 InChI Key: PWHULOQIROXLJO-UHFFFAOYSA-N Synonym: colloidal,mangan,manganese, elemental,cutaval,magnacat,metal alloy,tronamang,fume,mangan polish,compounds PubChem CID: 23930 ChEBI: CHEBI:35154 IUPAC Name: manganese SMILES: [Mn]
Manganese sputtering target, 76.2mm (3.0in) dia x 3.18mm (0.125in) thick, 99.95% (metals basis)
CAS: 7439-96-5 Molecular Formula: Mn Molecular Weight (g/mol): 54.94 MDL Number: MFCD00011111 InChI Key: PWHULOQIROXLJO-UHFFFAOYSA-N Synonym: colloidal,mangan,manganese, elemental,cutaval,magnacat,metal alloy,tronamang,fume,mangan polish,compounds PubChem CID: 23930 ChEBI: CHEBI:35154 IUPAC Name: manganese SMILES: [Mn]
Manganese Metal, Flakes, 99.5%, Spectrum™ Chemical
CAS: 7439-96-5 Molecular Weight (g/mol): 54.94
Manganese granules, 0.8-12mm (0.03-0.47in), 99.98% (metals basis)
CAS: 7439-96-5 Molecular Formula: Mn MDL Number: MFCD00011111 InChI Key: PWHULOQIROXLJO-UHFFFAOYSA-N Synonym: colloidal,mangan,manganese, elemental,cutaval,magnacat,metal alloy,tronamang,fume,mangan polish,compounds PubChem CID: 23930 ChEBI: CHEBI:35154 IUPAC Name: manganese SMILES: [Mn]
Manganese, 99+%, powder, -40 mesh
CAS: 7439-96-5 Molecular Formula: Mn Molecular Weight (g/mol): 54.94 MDL Number: MFCD00011111 InChI Key: PWHULOQIROXLJO-UHFFFAOYSA-N Synonym: colloidal,mangan,manganese, elemental,cutaval,magnacat,metal alloy,tronamang,fume,mangan polish,compounds PubChem CID: 23930 ChEBI: CHEBI:35154 IUPAC Name: manganese SMILES: [Mn]
Manganese powder, -325 mesh, 99.95% (metals basis)
CAS: 7439-96-5 Molecular Formula: Mn Molecular Weight (g/mol): 54.94 MDL Number: MFCD00011111 InChI Key: PWHULOQIROXLJO-UHFFFAOYSA-N Synonym: colloidal,mangan,manganese, elemental,cutaval,magnacat,metal alloy,tronamang,fume,mangan polish,compounds PubChem CID: 23930 ChEBI: CHEBI:35154 IUPAC Name: manganese SMILES: [Mn]
Manganese sputtering target, 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.95% (metals basis), Thermo Scientific™
CAS: 7439-96-5 Molecular Formula: Mn Molecular Weight (g/mol): 54.94 MDL Number: MFCD00011111 InChI Key: PWHULOQIROXLJO-UHFFFAOYSA-N Synonym: colloidal,mangan,manganese, elemental,cutaval,magnacat,metal alloy,tronamang,fume,mangan polish,compounds PubChem CID: 23930 ChEBI: CHEBI:35154 IUPAC Name: manganese SMILES: [Mn]
Manganese powder, APS <10 micron, 99.6% (metals basis)
CAS: 7439-96-5 Molecular Formula: Mn Molecular Weight (g/mol): 54.94 MDL Number: MFCD00011111 InChI Key: PWHULOQIROXLJO-UHFFFAOYSA-N Synonym: colloidal,mangan,manganese, elemental,cutaval,magnacat,metal alloy,tronamang,fume,mangan polish,compounds PubChem CID: 23930 ChEBI: CHEBI:35154 IUPAC Name: manganese SMILES: [Mn]