CAS RN 7439-97-6
Mercury, ACS, 99.999% (metals basis)
CAS: 7439-97-6 Molecular Formula: Hg Molecular Weight (g/mol): 200.59 MDL Number: MFCD00011035 InChI Key: QSHDDOUJBYECFT-UHFFFAOYSA-N Synonym: hydrargyrum,quicksilver,metallic,liquid silver,mercurio,quecksilber,mercure,quick silver,colloidal,mercury, metallic PubChem CID: 23931 ChEBI: CHEBI:16170 IUPAC Name: mercury SMILES: [Hg]
Mercury Standard For ICP, 10μg/mL in 5% HNO3, SPEX CertiPrep™
CAS: 7439-97-6 Molecular Formula: Hg Molecular Weight (g/mol): 200.59 MDL Number: MFCD00011035 InChI Key: QSHDDOUJBYECFT-UHFFFAOYSA-N PubChem CID: 23931 ChEBI: CHEBI:16170 IUPAC Name: mercury SMILES: [Hg]
Mercury Standard For ICP, 1000μg/mL, SPEX CertiPrep™
CAS: 7439-97-6 Molecular Formula: Hg Molecular Weight (g/mol): 200.59 MDL Number: MFCD00011035 InChI Key: QSHDDOUJBYECFT-UHFFFAOYSA-N PubChem CID: 23931 ChEBI: CHEBI:16170 IUPAC Name: mercury SMILES: [Hg]
Mercury, Electronic Grade, 99.9998% (metals basis)
CAS: 7439-97-6 Molecular Formula: Hg Molecular Weight (g/mol): 200.59 MDL Number: MFCD00011035 InChI Key: QSHDDOUJBYECFT-UHFFFAOYSA-N PubChem CID: 23931 ChEBI: CHEBI:16170 IUPAC Name: mercury SMILES: [Hg]
Mercury, plasma standard solution, Specpure™ Hg 1000μg/mL
CAS: 7439-97-6 Molecular Formula: Hg Molecular Weight (g/mol): 200.59 MDL Number: MFCD00011035 InChI Key: QSHDDOUJBYECFT-UHFFFAOYSA-N PubChem CID: 23931 ChEBI: CHEBI:16170 IUPAC Name: mercury SMILES: [Hg]
ICP-MS Mercury Single Element Standard, SPEX CertiPrep™
CAS: 7439-97-6 Molecular Formula: Hg Molecular Weight (g/mol): 200.59 MDL Number: MFCD00011035 InChI Key: QSHDDOUJBYECFT-UHFFFAOYSA-N PubChem CID: 23931 ChEBI: CHEBI:16170 IUPAC Name: mercury SMILES: [Hg]
Mercury, Triple Distilled, Avantor ANALYZED™ A.C.S. Reagent, Avantor™
CAS: 7439-97-6 Molecular Formula: Hg Molecular Weight (g/mol): 200.59 MDL Number: MFCD00011035 InChI Key: QSHDDOUJBYECFT-UHFFFAOYSA-N Synonym: hydrargyrum,quicksilver,metallic,liquid silver,mercurio,quecksilber,mercure,quick silver,colloidal,mercury, metallic PubChem CID: 23931 ChEBI: CHEBI:16170 IUPAC Name: mercury SMILES: [Hg]
Instrument Calibration Standard 6 For CLP, SPEX CertiPrep™
CAS: 7439-97-6 Molecular Formula: Hg Molecular Weight (g/mol): 200.59 MDL Number: MFCD00011035 InChI Key: QSHDDOUJBYECFT-UHFFFAOYSA-N PubChem CID: 23931 ChEBI: CHEBI:16170 IUPAC Name: mercury SMILES: [Hg]
Mercury ICP-MS Standard, 10,000 ppm Hg in 10% HNO3, Ricca Chemical
CAS: 7439-97-6 Molecular Formula: Hg Molecular Weight (g/mol): 200.59 MDL Number: MFCD00011035 InChI Key: QSHDDOUJBYECFT-UHFFFAOYSA-N PubChem CID: 23931 ChEBI: CHEBI:16170 IUPAC Name: mercury SMILES: [Hg]
Mercury ICP-MS Standard, 1000 ppm Hg in 3% HNO3, Ricca Chemical
CAS: 7439-97-6 Molecular Formula: Hg Molecular Weight (g/mol): 200.59 MDL Number: MFCD00011035 InChI Key: QSHDDOUJBYECFT-UHFFFAOYSA-N PubChem CID: 23931 ChEBI: CHEBI:16170 IUPAC Name: mercury SMILES: [Hg]
Mercury ICP Standard, 10,000 ppm Hg in 10% HNO3, Ricca Chemical
CAS: 7439-97-6 Molecular Formula: Hg Molecular Weight (g/mol): 200.59 MDL Number: MFCD00011035 InChI Key: QSHDDOUJBYECFT-UHFFFAOYSA-N PubChem CID: 23931 ChEBI: CHEBI:16170 IUPAC Name: mercury SMILES: [Hg]
Mercury ICP Standard, 1000 ppm Hg in 3% HNO3, Ricca Chemical
CAS: 7439-97-6 Molecular Formula: Hg Molecular Weight (g/mol): 200.59 MDL Number: MFCD00011035 InChI Key: QSHDDOUJBYECFT-UHFFFAOYSA-N PubChem CID: 23931 ChEBI: CHEBI:16170 IUPAC Name: mercury SMILES: [Hg]
Mercury Standard, 20 ppm Hg, Ricca Chemical
CAS: 7439-97-6 Molecular Formula: Hg Molecular Weight (g/mol): 200.59 MDL Number: MFCD00011035 InChI Key: QSHDDOUJBYECFT-UHFFFAOYSA-N PubChem CID: 23931 ChEBI: CHEBI:16170 IUPAC Name: mercury SMILES: [Hg]
Mercury GFAA Cal/Spike Standard - 100 ppm Hg in 5% HNO3, Ricca Chemical
CAS: 7439-97-6 Molecular Formula: Hg Molecular Weight (g/mol): 200.59 MDL Number: MFCD00011035 InChI Key: QSHDDOUJBYECFT-UHFFFAOYSA-N PubChem CID: 23931 ChEBI: CHEBI:16170 IUPAC Name: mercury SMILES: [Hg]
Mercury Standard For ICP, 100μg/mL in 5% HNO3, SPEX CertiPrep™
CAS: 7439-97-6 Molecular Formula: Hg Molecular Weight (g/mol): 200.59 MDL Number: MFCD00011035 InChI Key: QSHDDOUJBYECFT-UHFFFAOYSA-N PubChem CID: 23931 ChEBI: CHEBI:16170 IUPAC Name: mercury SMILES: [Hg]
- 1
- 2