CAS RN 7439-98-7
Molybdenum powder, -325 mesh, Puratronic™, 99.997% (metals basis excluding W), W <130ppm, Thermo Scientific Chemicals
CAS: 7439-98-7 Molecular Formula: Mo Molecular Weight (g/mol): 95.95 MDL Number: MFCD00003465 InChI Key: ZOKXTWBITQBERF-UHFFFAOYSA-N Synonym: molybdenum, elemental,6+ ion,mchvl,vi ion,molybdenum, metallic,metco 63,atom,molybdenum, ion 6+,tsm1,amperit 106.2 PubChem CID: 23932 ChEBI: CHEBI:28685 IUPAC Name: molybdenum SMILES: [Mo]
Molybdenum powder, APS 3-7 micron, 99.95% (metals basis), Thermo Scientific Chemicals
CAS: 7439-98-7 Molecular Formula: Mo Molecular Weight (g/mol): 95.95 MDL Number: MFCD00003465 InChI Key: ZOKXTWBITQBERF-UHFFFAOYSA-N Synonym: molybdenum, elemental,6+ ion,mchvl,vi ion,molybdenum, metallic,metco 63,atom,molybdenum, ion 6+,tsm1,amperit 106.2 PubChem CID: 23932 ChEBI: CHEBI:28685 IUPAC Name: molybdenum SMILES: [Mo]
Molybdenum foil, 0.025mm (0.001in) thick, 99.95% (metals basis)
CAS: 7439-98-7 Molecular Formula: Mo Molecular Weight (g/mol): 95.95 MDL Number: MFCD00003465 InChI Key: ZOKXTWBITQBERF-UHFFFAOYSA-N Synonym: molybdenum, elemental,6+ ion,mchvl,vi ion,molybdenum, metallic,metco 63,atom,molybdenum, ion 6+,tsm1,amperit 106.2 PubChem CID: 23932 ChEBI: CHEBI:28685 IUPAC Name: molybdenum SMILES: [Mo]
Molybdenum foil, 0.05mm (0.002in) thick, 99.95% (metals basis)
CAS: 7439-98-7 Molecular Formula: Mo Molecular Weight (g/mol): 95.95 MDL Number: MFCD00003465 InChI Key: ZOKXTWBITQBERF-UHFFFAOYSA-N Synonym: molybdenum, elemental,6+ ion,mchvl,vi ion,molybdenum, metallic,metco 63,atom,molybdenum, ion 6+,tsm1,amperit 106.2 PubChem CID: 23932 ChEBI: CHEBI:28685 IUPAC Name: molybdenum SMILES: [Mo]
Molybdenum powder, APS typically 2-4 micron, 99.9% (metals basis), Thermo Scientific Chemicals
CAS: 7439-98-7 Molecular Formula: Mo Molecular Weight (g/mol): 95.95 MDL Number: MFCD00003465 InChI Key: ZOKXTWBITQBERF-UHFFFAOYSA-N Synonym: molybdenum, elemental,6+ ion,mchvl,vi ion,molybdenum, metallic,metco 63,atom,molybdenum, ion 6+,tsm1,amperit 106.2 PubChem CID: 23932 ChEBI: CHEBI:28685 IUPAC Name: molybdenum SMILES: [Mo]
Molybdenum foil, 0.1mm (0.004in) thick, 99.95% (metals basis)
CAS: 7439-98-7 Molecular Formula: Mo Molecular Weight (g/mol): 95.95 MDL Number: MFCD00003465 InChI Key: ZOKXTWBITQBERF-UHFFFAOYSA-N Synonym: molybdenum, elemental,6+ ion,mchvl,vi ion,molybdenum, metallic,metco 63,atom,molybdenum, ion 6+,tsm1,amperit 106.2 PubChem CID: 23932 ChEBI: CHEBI:28685 IUPAC Name: molybdenum SMILES: [Mo]
Molybdenum, plasma standard solution, Specpure™ Mo 1000μg/mL
CAS: 7439-98-7 Molecular Formula: Mo Molecular Weight (g/mol): 95.95 MDL Number: MFCD00003465 InChI Key: ZOKXTWBITQBERF-UHFFFAOYSA-N PubChem CID: 23932 ChEBI: CHEBI:28685 IUPAC Name: molybdenum SMILES: [Mo]
Molybdenum powder, spherical, -170 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
CAS: 7439-98-7 Molecular Formula: Mo Molecular Weight (g/mol): 95.95 MDL Number: MFCD00003465 InChI Key: ZOKXTWBITQBERF-UHFFFAOYSA-N Synonym: molybdenum, elemental,6+ ion,mchvl,vi ion,molybdenum, metallic,metco 63,atom,molybdenum, ion 6+,tsm1,amperit 106.2 PubChem CID: 23932 ChEBI: CHEBI:28685 IUPAC Name: molybdenum SMILES: [Mo]
Molybdenum slug, 6.35mm (0.25 in.) dia. x 12.7mm (0.50 in.) length, 99.95% (metals basis), Thermo Scientific Chemicals
CAS: 7439-98-7 Molecular Formula: Mo Molecular Weight (g/mol): 95.95 MDL Number: MFCD00003465 InChI Key: ZOKXTWBITQBERF-UHFFFAOYSA-N Synonym: molybdenum, elemental,6+ ion,mchvl,vi ion,molybdenum, metallic,metco 63,atom,molybdenum, ion 6+,tsm1,amperit 106.2 PubChem CID: 23932 ChEBI: CHEBI:28685 IUPAC Name: molybdenum SMILES: [Mo]
Molybdenum pellet, 99.7% (metals basis)
CAS: 7439-98-7 Molecular Formula: Mo Molecular Weight (g/mol): 95.95 MDL Number: MFCD00003465 InChI Key: ZOKXTWBITQBERF-UHFFFAOYSA-N Synonym: molybdenum, elemental,6+ ion,mchvl,vi ion,molybdenum, metallic,metco 63,atom,molybdenum, ion 6+,tsm1,amperit 106.2 PubChem CID: 23932 ChEBI: CHEBI:28685 IUPAC Name: molybdenum SMILES: [Mo]
Molybdenum powder, -250 mesh, 99.9% (metals basis), Thermo Scientific Chemicals
CAS: 7439-98-7 Molecular Formula: Mo Molecular Weight (g/mol): 95.95 MDL Number: MFCD00003465 InChI Key: ZOKXTWBITQBERF-UHFFFAOYSA-N Synonym: molybdenum, elemental,6+ ion,mchvl,vi ion,molybdenum, metallic,metco 63,atom,molybdenum, ion 6+,tsm1,amperit 106.2 PubChem CID: 23932 ChEBI: CHEBI:28685 IUPAC Name: molybdenum SMILES: [Mo]
Molybdenum wire, platinum coated, 0.457mm (0.018in) dia
CAS: 7439-98-7 Molecular Formula: Mo Molecular Weight (g/mol): 95.95 MDL Number: MFCD00003465 InChI Key: ZOKXTWBITQBERF-UHFFFAOYSA-N Synonym: molybdenum, elemental,6+ ion,mchvl,vi ion,molybdenum, metallic,metco 63,atom,molybdenum, ion 6+,tsm1,amperit 106.2 PubChem CID: 23932 ChEBI: CHEBI:28685 IUPAC Name: molybdenum SMILES: [Mo]
Molybdenum foil, 0.127mm (0.005in) thick, 99.95% (metals basis)
CAS: 7439-98-7 Molecular Formula: Mo Molecular Weight (g/mol): 95.95 MDL Number: MFCD00003465 InChI Key: ZOKXTWBITQBERF-UHFFFAOYSA-N Synonym: molybdenum, elemental,6+ ion,mchvl,vi ion,molybdenum, metallic,metco 63,atom,molybdenum, ion 6+,tsm1,amperit 106.2 PubChem CID: 23932 ChEBI: CHEBI:28685 IUPAC Name: molybdenum SMILES: [Mo]
Molybdenum gauze, 165 mesh woven from 0.0509mm (0.002in) dia wire, twilled weave
CAS: 7439-98-7 Molecular Formula: Mo Molecular Weight (g/mol): 95.95 MDL Number: MFCD00003465 InChI Key: ZOKXTWBITQBERF-UHFFFAOYSA-N Synonym: molybdenum, elemental,6+ ion,mchvl,vi ion,molybdenum, metallic,metco 63,atom,molybdenum, ion 6+,tsm1,amperit 106.2 PubChem CID: 23932 ChEBI: CHEBI:28685 IUPAC Name: molybdenum SMILES: [Mo]
Molybdenum wire, 2.0mm (0.08in) dia, annealed, 99.95% (metals basis)
CAS: 7439-98-7 Molecular Formula: Mo Molecular Weight (g/mol): 95.95 MDL Number: MFCD00003465 InChI Key: ZOKXTWBITQBERF-UHFFFAOYSA-N Synonym: molybdenum, elemental,6+ ion,mchvl,vi ion,molybdenum, metallic,metco 63,atom,molybdenum, ion 6+,tsm1,amperit 106.2 PubChem CID: 23932 ChEBI: CHEBI:28685 IUPAC Name: molybdenum SMILES: [Mo]