CAS RN 7440-00-8
Neodymium powder, -200 mesh, 99.9% (REO)
CAS: 7440-00-8 Molecular Formula: Nd Molecular Weight (g/mol): 144.24 MDL Number: MFCD00011130 InChI Key: QEFYFXOXNSNQGX-UHFFFAOYSA-N Synonym: neodimio,neodyme,neodym,trihydride,ingot reo,neodymium, ion nd5+,atom,ingot,iii,foil, 3n PubChem CID: 23934 ChEBI: CHEBI:33372 IUPAC Name: neodymium SMILES: [Nd]
Neodymium rod, 12.7mm (0.5in) dia, 99.5% (metals basis excluding Ta)
CAS: 7440-00-8 Molecular Formula: Nd Molecular Weight (g/mol): 144.24 MDL Number: MFCD00011130 InChI Key: QEFYFXOXNSNQGX-UHFFFAOYSA-N Synonym: neodimio,neodyme,neodym,trihydride,ingot reo,neodymium, ion nd5+,atom,ingot,iii,foil, 3n PubChem CID: 23934 ChEBI: CHEBI:33372 IUPAC Name: neodymium SMILES: [Nd]
Neodymium foil, 0.1mm (0.004in) thick, 99.9% (REO)
CAS: 7440-00-8 Molecular Formula: Nd Molecular Weight (g/mol): 144.24 MDL Number: MFCD00011130 InChI Key: QEFYFXOXNSNQGX-UHFFFAOYSA-N Synonym: neodimio,neodyme,neodym,trihydride,ingot reo,neodymium, ion nd5+,atom,ingot,iii,foil, 3n PubChem CID: 23934 ChEBI: CHEBI:33372 IUPAC Name: neodymium SMILES: [Nd]
Neodymium foil, 0.3mm (0.012in) thick, 99.9% (REO)
CAS: 7440-00-8 Molecular Formula: Nd Molecular Weight (g/mol): 144.24 MDL Number: MFCD00011130 InChI Key: QEFYFXOXNSNQGX-UHFFFAOYSA-N Synonym: neodimio,neodyme,neodym,trihydride,ingot reo,neodymium, ion nd5+,atom,ingot,iii,foil, 3n PubChem CID: 23934 ChEBI: CHEBI:33372 IUPAC Name: neodymium SMILES: [Nd]
Neodymium powder, -40 mesh, 99.8% (REO)
CAS: 7440-00-8 Molecular Formula: Nd Molecular Weight (g/mol): 144.24 MDL Number: MFCD00011130 InChI Key: QEFYFXOXNSNQGX-UHFFFAOYSA-N Synonym: neodimio,neodyme,neodym,trihydride,ingot reo,neodymium, ion nd5+,atom,ingot,iii,foil, 3n PubChem CID: 23934 ChEBI: CHEBI:33372 IUPAC Name: neodymium SMILES: [Nd]
Neodymium rod, 6.35mm (0.25in) dia, 99.5% (metals basis excluding Ta)
CAS: 7440-00-8 Molecular Formula: Nd Molecular Weight (g/mol): 144.24 MDL Number: MFCD00011130 InChI Key: QEFYFXOXNSNQGX-UHFFFAOYSA-N Synonym: neodimio,neodyme,neodym,trihydride,ingot reo,neodymium, ion nd5+,atom,ingot,iii,foil, 3n PubChem CID: 23934 ChEBI: CHEBI:33372 IUPAC Name: neodymium SMILES: [Nd]
Neodymium foil, 1.0mm (0.04in) thick, 99.9% (REO)
CAS: 7440-00-8 Molecular Formula: Nd Molecular Weight (g/mol): 144.24 MDL Number: MFCD00011130 InChI Key: QEFYFXOXNSNQGX-UHFFFAOYSA-N Synonym: neodimio,neodyme,neodym,trihydride,ingot reo,neodymium, ion nd5+,atom,ingot,iii,foil, 3n PubChem CID: 23934 ChEBI: CHEBI:33372 IUPAC Name: neodymium SMILES: [Nd]
Neodymium ingot, 99.1% (metals basis)
CAS: 7440-00-8 Molecular Formula: Nd Molecular Weight (g/mol): 144.24 MDL Number: MFCD00011130 InChI Key: QEFYFXOXNSNQGX-UHFFFAOYSA-N Synonym: neodimio,neodyme,neodym,trihydride,ingot reo,neodymium, ion nd5+,atom,ingot,iii,foil, 3n PubChem CID: 23934 ChEBI: CHEBI:33372 IUPAC Name: neodymium SMILES: [Nd]
Neodymium Standard For ICP/MS, SPEX CertiPrep™
CAS: 7440-00-8 Molecular Formula: Nd Molecular Weight (g/mol): 144.24 MDL Number: MFCD00011130 InChI Key: QEFYFXOXNSNQGX-UHFFFAOYSA-N PubChem CID: 23934 ChEBI: CHEBI:33372 IUPAC Name: neodymium SMILES: [Nd]
Neodymium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™
CAS: 7440-00-8 Molecular Formula: Nd Molecular Weight (g/mol): 144.24 MDL Number: MFCD00011130 InChI Key: QEFYFXOXNSNQGX-UHFFFAOYSA-N PubChem CID: 23934 ChEBI: CHEBI:33372 IUPAC Name: neodymium SMILES: [Nd]
Sigma Aldrich Neodymium
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Neodymium
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Neodymium
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Neodymium
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Neodymium
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
- 1
- 2