accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7440-10-0

IUPAC Name: praseodymium
Synonyms: praseodymium|praseodymium atom
Molecular Weight (g/mol): 140.91
Molecular Formula: Pr
InChi Key: PUDIUYLPXJFUGB-UHFFFAOYSA-N
SMILES: [Pr]
Quantity 
  • (20)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (4)
  • (2)
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (18)
  • (8)
  • (7)
Form 
  • (4)
  • (6)
  • (3)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (20)
  • (2)
  • (19)
  • (20)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (4)
  • (2)
  • (1)
  • (2)
  • (2)
  • (18)
  • (8)
  • (7)
  • (4)
  • (6)
  • (3)
  • (4)
  • (2)
  • (2)
  • (2)