CAS RN 7440-15-5
Rhenium powder, -22 mesh, Puratronic™, 99.997% (metals basis)
CAS: 7440-15-5 Molecular Formula: Re Molecular Weight (g/mol): 186.21 MDL Number: MFCD00011195 InChI Key: WUAPFZMCVAUBPE-UHFFFAOYSA-N Synonym: rhenium, elementar,renio,unii-7yhu292iny,7yhu292iny,foil,ion re7,rhenium, ion re7,atom,hydride,pellets PubChem CID: 23947 ChEBI: CHEBI:49882 IUPAC Name: rhenium SMILES: [Re]
Rhenium wire, 1.0mm (0.04in) dia, 99.97% (metals basis)
CAS: 7440-15-5 Molecular Formula: Re Molecular Weight (g/mol): 186.21 MDL Number: MFCD00011195 InChI Key: WUAPFZMCVAUBPE-UHFFFAOYSA-N Synonym: rhenium, elementar,renio,unii-7yhu292iny,7yhu292iny,foil,ion re7,rhenium, ion re7,atom,hydride,pellets PubChem CID: 23947 ChEBI: CHEBI:49882 IUPAC Name: rhenium SMILES: [Re]
Rhenium wire, 0.5mm (0.02in) dia, 99.97% (metals basis)
CAS: 7440-15-5 Molecular Formula: Re Molecular Weight (g/mol): 186.21 MDL Number: MFCD00011195 InChI Key: WUAPFZMCVAUBPE-UHFFFAOYSA-N Synonym: rhenium, elementar,renio,unii-7yhu292iny,7yhu292iny,foil,ion re7,rhenium, ion re7,atom,hydride,pellets PubChem CID: 23947 ChEBI: CHEBI:49882 IUPAC Name: rhenium SMILES: [Re]
Rhenium foil, 1.0mm (0.04in) thick, 99.97% (metals basis)
CAS: 7440-15-5 Molecular Formula: Re Molecular Weight (g/mol): 186.21 MDL Number: MFCD00011195 InChI Key: WUAPFZMCVAUBPE-UHFFFAOYSA-N Synonym: rhenium, elementar,renio,unii-7yhu292iny,7yhu292iny,foil,ion re7,rhenium, ion re7,atom,hydride,pellets PubChem CID: 23947 ChEBI: CHEBI:49882 IUPAC Name: rhenium SMILES: [Re]
Rhenium foil, 0.5mm (0.02in) thick, annealed, 99.97% (metals basis)
CAS: 7440-15-5 Molecular Formula: Re Molecular Weight (g/mol): 186.21 MDL Number: MFCD00011195 InChI Key: WUAPFZMCVAUBPE-UHFFFAOYSA-N Synonym: rhenium, elementar,renio,unii-7yhu292iny,7yhu292iny,foil,ion re7,rhenium, ion re7,atom,hydride,pellets PubChem CID: 23947 ChEBI: CHEBI:49882 IUPAC Name: rhenium SMILES: [Re]
Rhenium foil, 0.025mm (0.001in) thick, 99.97% (metals basis)
CAS: 7440-15-5 Molecular Formula: Re Molecular Weight (g/mol): 186.21 MDL Number: MFCD00011195 InChI Key: WUAPFZMCVAUBPE-UHFFFAOYSA-N Synonym: rhenium, elementar,renio,unii-7yhu292iny,7yhu292iny,foil,ion re7,rhenium, ion re7,atom,hydride,pellets PubChem CID: 23947 ChEBI: CHEBI:49882 IUPAC Name: rhenium SMILES: [Re]
Rhenium foil, 0.25mm (0.01in) thick, 99.97% (metals basis)
CAS: 7440-15-5 Molecular Formula: Re Molecular Weight (g/mol): 186.21 MDL Number: MFCD00011195 InChI Key: WUAPFZMCVAUBPE-UHFFFAOYSA-N Synonym: rhenium, elementar,renio,unii-7yhu292iny,7yhu292iny,foil,ion re7,rhenium, ion re7,atom,hydride,pellets PubChem CID: 23947 ChEBI: CHEBI:49882 IUPAC Name: rhenium SMILES: [Re]
Rhenium slug, 12.7mm (0.5 in.) dia., 99.99% (metals basis), Thermo Scientific Chemicals
CAS: 7440-15-5 Molecular Formula: Re Molecular Weight (g/mol): 186.21 MDL Number: MFCD00011195 InChI Key: WUAPFZMCVAUBPE-UHFFFAOYSA-N Synonym: rhenium, elementar,renio,unii-7yhu292iny,7yhu292iny,foil,ion re7,rhenium, ion re7,atom,hydride,pellets PubChem CID: 23947 ChEBI: CHEBI:49882 IUPAC Name: rhenium SMILES: [Re]
Rhenium foil, 0.1mm (0.004in) thick, 99.97% (metals basis)
CAS: 7440-15-5 Molecular Formula: Re Molecular Weight (g/mol): 186.21 MDL Number: MFCD00011195 InChI Key: WUAPFZMCVAUBPE-UHFFFAOYSA-N Synonym: rhenium, elementar,renio,unii-7yhu292iny,7yhu292iny,foil,ion re7,rhenium, ion re7,atom,hydride,pellets PubChem CID: 23947 ChEBI: CHEBI:49882 IUPAC Name: rhenium SMILES: [Re]
Rhenium, plasma standard solution, Specpure™ Re 1000μg/mL
CAS: 7440-15-5 Molecular Formula: Re Molecular Weight (g/mol): 186.21 MDL Number: MFCD00011195 InChI Key: WUAPFZMCVAUBPE-UHFFFAOYSA-N PubChem CID: 23947 ChEBI: CHEBI:49882 IUPAC Name: rhenium SMILES: [Re]
Rhenium powder, -325 mesh, 99.99% (metals basis)
CAS: 7440-15-5 Molecular Formula: Re Molecular Weight (g/mol): 186.21 MDL Number: MFCD00011195 InChI Key: WUAPFZMCVAUBPE-UHFFFAOYSA-N PubChem CID: 23947 ChEBI: CHEBI:49882 IUPAC Name: rhenium SMILES: [Re]
Rhenium wire, 0.1mm (0.004 in.) dia., annealed, 99.97% (metals basis), Thermo Scientific Chemicals
CAS: 7440-15-5 Molecular Formula: Re Molecular Weight (g/mol): 186.21 MDL Number: MFCD00011195 InChI Key: WUAPFZMCVAUBPE-UHFFFAOYSA-N Synonym: rhenium, elementar,renio,unii-7yhu292iny,7yhu292iny,foil,ion re7,rhenium, ion re7,atom,hydride,pellets PubChem CID: 23947 ChEBI: CHEBI:49882 IUPAC Name: rhenium SMILES: [Re]
Rhenium wire, 0.25mm (0.01in) dia, 99.97% (metals basis)
CAS: 7440-15-5 Molecular Formula: Re Molecular Weight (g/mol): 186.21 MDL Number: MFCD00011195 InChI Key: WUAPFZMCVAUBPE-UHFFFAOYSA-N Synonym: rhenium, elementar,renio,unii-7yhu292iny,7yhu292iny,foil,ion re7,rhenium, ion re7,atom,hydride,pellets PubChem CID: 23947 ChEBI: CHEBI:49882 IUPAC Name: rhenium SMILES: [Re]
Rhenium Standard For ICP, 1000μg/mL in H2O, Assurance Grade, SPEX CertiPrep™
CAS: 7440-15-5 Molecular Formula: Re Molecular Weight (g/mol): 186.21 MDL Number: MFCD00011195 InChI Key: WUAPFZMCVAUBPE-UHFFFAOYSA-N PubChem CID: 23947 ChEBI: CHEBI:49882 IUPAC Name: rhenium SMILES: [Re]
Rhenium Standard For ICP, 1000μg/mL in H2O, SPEX CertiPrep™
CAS: 7440-15-5 Molecular Formula: Re Molecular Weight (g/mol): 186.21 MDL Number: MFCD00011195 InChI Key: WUAPFZMCVAUBPE-UHFFFAOYSA-N PubChem CID: 23947 ChEBI: CHEBI:49882 IUPAC Name: rhenium SMILES: [Re]