CAS RN 7440-19-9
Samarium foil, 0.62mm (0.024in) thick, 99.9% (REO)
CAS: 7440-19-9 Molecular Formula: Sm Molecular Weight (g/mol): 150.36 MDL Number: MFCD00011233 MFCD00151299 InChI Key: KZUNJOHGWZRPMI-UHFFFAOYSA-N Synonym: samario,unii-42od65l39f,samarium, elemental,atom,powder,ingot,foil, 3n,chips, 3n,ingot, 3n,acmc-1bk0l PubChem CID: 23951 ChEBI: CHEBI:33374 IUPAC Name: samarium SMILES: [Sm]
Samarium ingot, 99.9% (REO)
CAS: 7440-19-9 Molecular Formula: Sm Molecular Weight (g/mol): 150.36 MDL Number: MFCD00011233 MFCD00151299 InChI Key: KZUNJOHGWZRPMI-UHFFFAOYSA-N Synonym: samario,unii-42od65l39f,samarium, elemental,atom,powder,ingot,foil, 3n,chips, 3n,ingot, 3n,acmc-1bk0l PubChem CID: 23951 ChEBI: CHEBI:33374 IUPAC Name: samarium SMILES: [Sm]
Samarium foil, 0.75mm (0.03 in.) thick, 99.9% (REO), Thermo Scientific Chemicals
CAS: 7440-19-9 Molecular Formula: Sm Molecular Weight (g/mol): 150.36 MDL Number: MFCD00011233 MFCD00151299 InChI Key: KZUNJOHGWZRPMI-UHFFFAOYSA-N Synonym: samario,unii-42od65l39f,samarium, elemental,atom,powder,ingot,foil, 3n,chips, 3n,ingot, 3n,acmc-1bk0l PubChem CID: 23951 ChEBI: CHEBI:33374 IUPAC Name: samarium SMILES: [Sm]
Samarium foil, 0.127mm (0.005in) thick, 99.9% (REO)
CAS: 7440-19-9 Molecular Formula: Sm Molecular Weight (g/mol): 150.36 MDL Number: MFCD00011233 MFCD00151299 InChI Key: KZUNJOHGWZRPMI-UHFFFAOYSA-N Synonym: samario,unii-42od65l39f,samarium, elemental,atom,powder,ingot,foil, 3n,chips, 3n,ingot, 3n,acmc-1bk0l PubChem CID: 23951 ChEBI: CHEBI:33374 IUPAC Name: samarium SMILES: [Sm]
Samarium foil, 0.25mm (0.01in) thick, 99.9% (REO)
CAS: 7440-19-9 Molecular Formula: Sm Molecular Weight (g/mol): 150.36 MDL Number: MFCD00011233 MFCD00151299 InChI Key: KZUNJOHGWZRPMI-UHFFFAOYSA-N Synonym: samario,unii-42od65l39f,samarium, elemental,atom,powder,ingot,foil, 3n,chips, 3n,ingot, 3n,acmc-1bk0l PubChem CID: 23951 ChEBI: CHEBI:33374 IUPAC Name: samarium SMILES: [Sm]
Samarium rod, 12.7mm (0.5in) dia, 99.9% (metals basis excluding Ta)
CAS: 7440-19-9 Molecular Formula: Sm Molecular Weight (g/mol): 150.36 MDL Number: MFCD00011233 MFCD00151299 InChI Key: KZUNJOHGWZRPMI-UHFFFAOYSA-N Synonym: samario,unii-42od65l39f,samarium, elemental,atom,powder,ingot,foil, 3n,chips, 3n,ingot, 3n,acmc-1bk0l PubChem CID: 23951 ChEBI: CHEBI:33374 IUPAC Name: samarium SMILES: [Sm]
Samarium, 99.9%, (trace metal basis), 20 to 40 mesh
CAS: 7440-19-9 Molecular Formula: Sm Molecular Weight (g/mol): 150.36 MDL Number: MFCD00011233 MFCD00151299 InChI Key: KZUNJOHGWZRPMI-UHFFFAOYSA-N Synonym: samario,unii-42od65l39f,samarium, elemental,atom,powder,ingot,foil, 3n,chips, 3n,ingot, 3n,acmc-1bk0l PubChem CID: 23951 ChEBI: CHEBI:33374 IUPAC Name: samarium SMILES: [Sm]
Samarium powder, -40 mesh, 99.9% (REO)
CAS: 7440-19-9 Molecular Formula: Sm Molecular Weight (g/mol): 150.36 MDL Number: MFCD00011233 MFCD00151299 InChI Key: KZUNJOHGWZRPMI-UHFFFAOYSA-N Synonym: samario,unii-42od65l39f,samarium, elemental,atom,powder,ingot,foil, 3n,chips, 3n,ingot, 3n,acmc-1bk0l PubChem CID: 23951 ChEBI: CHEBI:33374 IUPAC Name: samarium SMILES: [Sm]
Samarium ingot/button,≈50.8mm (2.0 in.) dia., 99.5% (REO)
CAS: 7440-19-9 Molecular Formula: Sm Molecular Weight (g/mol): 150.36 MDL Number: MFCD00011233 MFCD00151299 InChI Key: KZUNJOHGWZRPMI-UHFFFAOYSA-N Synonym: samario,unii-42od65l39f,samarium, elemental,atom,powder,ingot,foil, 3n,chips, 3n,ingot, 3n,acmc-1bk0l PubChem CID: 23951 ChEBI: CHEBI:33374 IUPAC Name: samarium SMILES: [Sm]
Samarium foil, 0.025mm (0.001in) thick, 99.9% (REO)
CAS: 7440-19-9 Molecular Formula: Sm Molecular Weight (g/mol): 150.36 MDL Number: MFCD00011233 MFCD00151299 InChI Key: KZUNJOHGWZRPMI-UHFFFAOYSA-N Synonym: samario,unii-42od65l39f,samarium, elemental,atom,powder,ingot,foil, 3n,chips, 3n,ingot, 3n,acmc-1bk0l PubChem CID: 23951 ChEBI: CHEBI:33374 IUPAC Name: samarium SMILES: [Sm]
Samarium rod, 6.35mm (0.25in) dia, 99.9% (metals basis excluding Ta)
CAS: 7440-19-9 Molecular Formula: Sm Molecular Weight (g/mol): 150.36 MDL Number: MFCD00011233 MFCD00151299 InChI Key: KZUNJOHGWZRPMI-UHFFFAOYSA-N Synonym: samario,unii-42od65l39f,samarium, elemental,atom,powder,ingot,foil, 3n,chips, 3n,ingot, 3n,acmc-1bk0l PubChem CID: 23951 ChEBI: CHEBI:33374 IUPAC Name: samarium SMILES: [Sm]
Samarium foil, 1.0mm (0.04in) thick, 99.9% (REO)
CAS: 7440-19-9 Molecular Formula: Sm Molecular Weight (g/mol): 150.36 MDL Number: MFCD00011233 MFCD00151299 InChI Key: KZUNJOHGWZRPMI-UHFFFAOYSA-N Synonym: samario,unii-42od65l39f,samarium, elemental,atom,powder,ingot,foil, 3n,chips, 3n,ingot, 3n,acmc-1bk0l PubChem CID: 23951 ChEBI: CHEBI:33374 IUPAC Name: samarium SMILES: [Sm]
Samarium pieces, sublimed dendritic, 99.9% (REO)
CAS: 7440-19-9 Molecular Formula: Sm Molecular Weight (g/mol): 150.36 MDL Number: MFCD00011233 MFCD00151299 InChI Key: KZUNJOHGWZRPMI-UHFFFAOYSA-N Synonym: samario,unii-42od65l39f,samarium, elemental,atom,powder,ingot,foil, 3n,chips, 3n,ingot, 3n,acmc-1bk0l PubChem CID: 23951 ChEBI: CHEBI:33374 IUPAC Name: samarium SMILES: [Sm]
Samarium foil, 0.1mm (0.004in) thick, 99.9% (REO)
CAS: 7440-19-9 Molecular Formula: Sm Molecular Weight (g/mol): 150.36 MDL Number: MFCD00011233 MFCD00151299 InChI Key: KZUNJOHGWZRPMI-UHFFFAOYSA-N Synonym: samario,unii-42od65l39f,samarium, elemental,atom,powder,ingot,foil, 3n,chips, 3n,ingot, 3n,acmc-1bk0l PubChem CID: 23951 ChEBI: CHEBI:33374 IUPAC Name: samarium SMILES: [Sm]
Samarium pieces, 12mm (0.47in) & down, 99.9% (REO)
CAS: 7440-19-9 Molecular Formula: Sm Molecular Weight (g/mol): 150.36 MDL Number: MFCD00011233 MFCD00151299 InChI Key: KZUNJOHGWZRPMI-UHFFFAOYSA-N Synonym: samario,unii-42od65l39f,samarium, elemental,atom,powder,ingot,foil, 3n,chips, 3n,ingot, 3n,acmc-1bk0l PubChem CID: 23951 ChEBI: CHEBI:33374 IUPAC Name: samarium SMILES: [Sm]