CAS RN 7440-23-5
Sodium ingot, 99.8% (metals basis)
CAS: 7440-23-5 Molecular Formula: Na Molecular Weight (g/mol): 22.99 MDL Number: MFCD00085307 InChI Key: KEAYESYHFKHZAL-UHFFFAOYSA-N Synonym: natrium,sodium-23,sodio,metal,sodio spanish,liquid alloy,unii-9nez333n27,hsdb 687,unii-23j3bhr95o,ingot PubChem CID: 5360545 ChEBI: CHEBI:26708 IUPAC Name: sodium SMILES: [Na]
Sodium, 99.8%, oiled sticks, wrapped in aluminum foil
CAS: 7440-23-5 Molecular Formula: Na Molecular Weight (g/mol): 22.99 MDL Number: MFCD00085307 InChI Key: KEAYESYHFKHZAL-UHFFFAOYSA-N Synonym: natrium,sodium-23,sodio,metal,sodio spanish,liquid alloy,unii-9nez333n27,hsdb 687,unii-23j3bhr95o,ingot PubChem CID: 5360545 ChEBI: CHEBI:26708 IUPAC Name: sodium SMILES: [Na]
Sodium, Ion chromatography standard solution, Specpure™ Na∣+ 1000μg/mL
CAS: 7440-23-5 Molecular Formula: Na Molecular Weight (g/mol): 22.99 MDL Number: MFCD00085307 InChI Key: KEAYESYHFKHZAL-UHFFFAOYSA-N Synonym: natrium,sodium-23,sodio,metal,sodio spanish,liquid alloy,unii-9nez333n27,hsdb 687,unii-23j3bhr95o,monosodium PubChem CID: 5360545 ChEBI: CHEBI:26708 IUPAC Name: sodium SMILES: [Na]
Sodium, 30wt% dispersion in toluene, <0.1 mm particle size
CAS: 7440-23-5 Molecular Formula: Na Molecular Weight (g/mol): 22.99 MDL Number: MFCD00085307 InChI Key: KEAYESYHFKHZAL-UHFFFAOYSA-N Synonym: natrium,sodium-23,sodio,metal,sodio spanish,liquid alloy,unii-9nez333n27,hsdb 687,unii-23j3bhr95o,ingot PubChem CID: 5360545 ChEBI: CHEBI:26708 IUPAC Name: sodium SMILES: [Na]
Sodium, 99.95% (metals basis)
CAS: 7440-23-5 Molecular Formula: Na Molecular Weight (g/mol): 22.99 MDL Number: MFCD00085307 InChI Key: KEAYESYHFKHZAL-UHFFFAOYSA-N Synonym: natrium,sodium-23,sodio,metal,sodio spanish,liquid alloy,unii-9nez333n27,hsdb 687,unii-23j3bhr95o,ingot PubChem CID: 5360545 ChEBI: CHEBI:26708 IUPAC Name: sodium SMILES: [Na]
Sodium sticks, coated in film of protective hydrocarbon oil, 99%
CAS: 7440-23-5 Molecular Formula: Na Molecular Weight (g/mol): 22.99 MDL Number: MFCD00085307 InChI Key: KEAYESYHFKHZAL-UHFFFAOYSA-N Synonym: natrium,sodium-23,sodio,metal,sodio spanish,liquid alloy,unii-9nez333n27,hsdb 687,unii-23j3bhr95o,ingot PubChem CID: 5360545 ChEBI: CHEBI:26708 IUPAC Name: sodium SMILES: [Na]
Sodium, dispersion, 40% in oil
CAS: 7440-23-5 Molecular Formula: Na Molecular Weight (g/mol): 22.99 MDL Number: MFCD00085307 InChI Key: KEAYESYHFKHZAL-UHFFFAOYSA-N Synonym: natrium,sodium-23,sodio,metal,sodio spanish,liquid alloy,unii-9nez333n27,hsdb 687,unii-23j3bhr95o,ingot PubChem CID: 5360545 ChEBI: CHEBI:26708 IUPAC Name: sodium SMILES: [Na]
Sodium, dry stick
CAS: 7440-23-5 Molecular Formula: Na Molecular Weight (g/mol): 22.99 MDL Number: MFCD00085307 InChI Key: KEAYESYHFKHZAL-UHFFFAOYSA-N Synonym: natrium,sodium-23,sodio,metal,sodio spanish,liquid alloy,unii-9nez333n27,hsdb 687,unii-23j3bhr95o,ingot PubChem CID: 5360545 ChEBI: CHEBI:26708 IUPAC Name: sodium SMILES: [Na]
Sodium Standard For ICP, 1000μg/mL in 2% HCl, SPEX CertiPrep™
CAS: 7440-23-5 Molecular Formula: Na Molecular Weight (g/mol): 22.99 MDL Number: MFCD00085307 InChI Key: KEAYESYHFKHZAL-UHFFFAOYSA-N PubChem CID: 5360545 ChEBI: CHEBI:26708 IUPAC Name: sodium SMILES: [Na]
Sodium Standard For ICP, 10,000μg/mL in 5% HNO3, SPEX CertiPrep™
CAS: 7440-23-5 Molecular Formula: Na Molecular Weight (g/mol): 22.99 MDL Number: MFCD00085307 InChI Key: KEAYESYHFKHZAL-UHFFFAOYSA-N PubChem CID: 5360545 ChEBI: CHEBI:26708 IUPAC Name: sodium SMILES: [Na]
Sodium Standard For ICP/MS, Claritas PPT Grade, SPEX CertiPrep™
CAS: 7440-23-5 Molecular Formula: Na Molecular Weight (g/mol): 22.99 MDL Number: MFCD00085307 InChI Key: KEAYESYHFKHZAL-UHFFFAOYSA-N PubChem CID: 5360545 ChEBI: CHEBI:26708 IUPAC Name: sodium SMILES: [Na]
Sodium Standard Single Element Ion Standard - Cation, SPEX CertiPrep™
CAS: 7440-23-5 Molecular Formula: Na Molecular Weight (g/mol): 22.99 MDL Number: MFCD00085307 InChI Key: KEAYESYHFKHZAL-UHFFFAOYSA-N PubChem CID: 5360545 ChEBI: CHEBI:26708 IUPAC Name: sodium SMILES: [Na]
Sodium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Sodium Standard in Base Oil 75, 5000μg/g, SPEX CertiPrep™
CAS: 7440-23-5 Molecular Formula: Na Molecular Weight (g/mol): 22.99 MDL Number: MFCD00085307 InChI Key: KEAYESYHFKHZAL-UHFFFAOYSA-N PubChem CID: 5360545 ChEBI: CHEBI:26708 IUPAC Name: sodium SMILES: [Na]
- 1
- 2