CAS RN 7440-25-7
Tantalum powder, APS ^=2 micron, 99.9% (metals basis)
CAS: 7440-25-7 Molecular Formula: Ta Molecular Weight (g/mol): 180.95 MDL Number: MFCD00011252 InChI Key: GUVRBAGPIYLISA-UHFFFAOYSA-N Synonym: tantalum, elemental,hydride,tantalum-181,tantalum, metal and oxide dust,tantal,tantalum, metal,powder,foil,wire,rod PubChem CID: 23956 ChEBI: CHEBI:33348 IUPAC Name: tantalum SMILES: [Ta]
Tantalum powder, -325 mesh, 99.9% (metals basis), 99.6%
CAS: 7440-25-7 Molecular Formula: Ta Molecular Weight (g/mol): 180.95 MDL Number: MFCD00011252 InChI Key: GUVRBAGPIYLISA-UHFFFAOYSA-N Synonym: tantalum, elemental,hydride,tantalum-181,tantalum, metal and oxide dust,tantal,tantalum, metal,powder,foil,wire,rod PubChem CID: 23956 ChEBI: CHEBI:33348 IUPAC Name: tantalum SMILES: [Ta]
Tantalum foil, 0.025mm (0.001in) thick, 99.997% (metals basis)
CAS: 7440-25-7 Molecular Formula: Ta Molecular Weight (g/mol): 180.95 MDL Number: MFCD00011252 InChI Key: GUVRBAGPIYLISA-UHFFFAOYSA-N Synonym: tantalum, elemental,hydride,tantalum-181,tantalum, metal and oxide dust,tantal,tantalum, metal,powder,foil,wire,rod PubChem CID: 23956 ChEBI: CHEBI:33348 IUPAC Name: tantalum SMILES: [Ta]
Tantalum rod, 4.75mm (0.19in) dia, 99.9% (metals basis)
CAS: 7440-25-7 Molecular Formula: Ta Molecular Weight (g/mol): 180.95 MDL Number: MFCD00011252 InChI Key: GUVRBAGPIYLISA-UHFFFAOYSA-N Synonym: tantalum, elemental,hydride,tantalum-181,tantalum, metal and oxide dust,tantal,tantalum, metal,powder,foil,wire,rod PubChem CID: 23956 ChEBI: CHEBI:33348 IUPAC Name: tantalum SMILES: [Ta]
Tantalum foil, 0.025mm (0.001in) thick, hard, 99.95% (metals basis)
CAS: 7440-25-7 Molecular Formula: Ta Molecular Weight (g/mol): 180.95 MDL Number: MFCD00011252 InChI Key: GUVRBAGPIYLISA-UHFFFAOYSA-N Synonym: tantalum, elemental,hydride,tantalum-181,tantalum, metal and oxide dust,tantal,tantalum, metal,powder,foil,wire,rod PubChem CID: 23956 ChEBI: CHEBI:33348 IUPAC Name: tantalum SMILES: [Ta]
Tantalum wire, 1.0mm (0.04in) dia, annealed, 99.95% (metals basis)
CAS: 7440-25-7 Molecular Formula: Ta Molecular Weight (g/mol): 180.95 MDL Number: MFCD00011252 InChI Key: GUVRBAGPIYLISA-UHFFFAOYSA-N Synonym: tantalum, elemental,hydride,tantalum-181,tantalum, metal and oxide dust,tantal,tantalum, metal,powder,foil,wire,rod PubChem CID: 23956 ChEBI: CHEBI:33348 IUPAC Name: tantalum SMILES: [Ta]
Tantalum wire, 1.5mm (0.06in) dia, annealed, 99.95% (metals basis)
CAS: 7440-25-7 Molecular Formula: Ta Molecular Weight (g/mol): 180.95 MDL Number: MFCD00011252 InChI Key: GUVRBAGPIYLISA-UHFFFAOYSA-N Synonym: tantalum, elemental,hydride,tantalum-181,tantalum, metal and oxide dust,tantal,tantalum, metal,powder,foil,wire,rod PubChem CID: 23956 ChEBI: CHEBI:33348 IUPAC Name: tantalum SMILES: [Ta]
Tantalum foil, 2.0mm (0.08in) thick, 99.95% (metals basis)
CAS: 7440-25-7 Molecular Formula: Ta Molecular Weight (g/mol): 180.95 MDL Number: MFCD00011252 InChI Key: GUVRBAGPIYLISA-UHFFFAOYSA-N Synonym: tantalum, elemental,hydride,tantalum-181,tantalum, metal and oxide dust,tantal,tantalum, metal,powder,foil,wire,rod PubChem CID: 23956 ChEBI: CHEBI:33348 IUPAC Name: tantalum SMILES: [Ta]
Tantalum rod, 4.5mm (0.18 in.) dia., Puratronic™, 99.96% (metals basis), Nb nominal 50ppm, Thermo Scientific Chemicals
CAS: 7440-25-7 Molecular Formula: Ta Molecular Weight (g/mol): 180.95 MDL Number: MFCD00011252 InChI Key: GUVRBAGPIYLISA-UHFFFAOYSA-N Synonym: tantalum, elemental,hydride,tantalum-181,tantalum, metal and oxide dust,tantal,tantalum, metal,powder,foil,wire,rod PubChem CID: 23956 ChEBI: CHEBI:33348 IUPAC Name: tantalum SMILES: [Ta]
Tantalum Thinfoil, 0.0025mm (0.0001in) thick, 99.9% (metals basis)
CAS: 7440-25-7 Molecular Formula: Ta Molecular Weight (g/mol): 180.95 MDL Number: MFCD00011252 InChI Key: GUVRBAGPIYLISA-UHFFFAOYSA-N Synonym: tantalum, elemental,hydride,tantalum-181,tantalum, metal and oxide dust,tantal,tantalum, metal,powder,foil,wire,rod PubChem CID: 23956 ChEBI: CHEBI:33348 IUPAC Name: tantalum SMILES: [Ta]
Tantalum wire, 0.05mm (0.002in) dia, annealed, 99.9% (metals basis)
CAS: 7440-25-7 Molecular Formula: Ta Molecular Weight (g/mol): 180.95 MDL Number: MFCD00011252 InChI Key: GUVRBAGPIYLISA-UHFFFAOYSA-N Synonym: tantalum, elemental,hydride,tantalum-181,tantalum, metal and oxide dust,tantal,tantalum, metal,powder,foil,wire,rod PubChem CID: 23956 ChEBI: CHEBI:33348 IUPAC Name: tantalum SMILES: [Ta]
Tantalum slug, 3.175mm (0.125in) dia x 3.175mm (0.125in) length, 99.95% (metals basis)
CAS: 7440-25-7 Molecular Formula: Ta Molecular Weight (g/mol): 180.95 MDL Number: MFCD00011252 InChI Key: GUVRBAGPIYLISA-UHFFFAOYSA-N Synonym: tantalum, elemental,hydride,tantalum-181,tantalum, metal and oxide dust,tantal,tantalum, metal,powder,foil,wire,rod PubChem CID: 23956 ChEBI: CHEBI:33348 IUPAC Name: tantalum SMILES: [Ta]
Tantalum gauze, 80 mesh woven from 0.08mm (0.003 in.) dia. wire, Thermo Scientific Chemicals
CAS: 7440-25-7 Molecular Formula: Ta Molecular Weight (g/mol): 180.95 MDL Number: MFCD00011252 InChI Key: GUVRBAGPIYLISA-UHFFFAOYSA-N Synonym: tantalum, elemental,hydride,tantalum-181,tantalum, metal and oxide dust,tantal,tantalum, metal,powder,foil,wire,rod PubChem CID: 23956 ChEBI: CHEBI:33348 IUPAC Name: tantalum SMILES: [Ta]
Tantalum slug, 6.35mm (0.25in) dia x 6.35mm (0.25in) length, 99.95% (metals basis)
CAS: 7440-25-7 Molecular Formula: Ta Molecular Weight (g/mol): 180.95 MDL Number: MFCD00011252 InChI Key: GUVRBAGPIYLISA-UHFFFAOYSA-N Synonym: tantalum, elemental,hydride,tantalum-181,tantalum, metal and oxide dust,tantal,tantalum, metal,powder,foil,wire,rod PubChem CID: 23956 ChEBI: CHEBI:33348 IUPAC Name: tantalum SMILES: [Ta]
Tantalum pellet, 1mm (0.04in) dia x 2mm (0.08in), 99.9% (metals basis)
CAS: 7440-25-7 Molecular Formula: Ta Molecular Weight (g/mol): 180.95 MDL Number: MFCD00011252 InChI Key: GUVRBAGPIYLISA-UHFFFAOYSA-N Synonym: tantalum, elemental,hydride,tantalum-181,tantalum, metal and oxide dust,tantal,tantalum, metal,powder,foil,wire,rod PubChem CID: 23956 ChEBI: CHEBI:33348 IUPAC Name: tantalum SMILES: [Ta]