accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7440-29-1

IUPAC Name: thorium
Synonyms: thorium torio
Molecular Weight (g/mol): 232.04
Molecular Formula: Th
InChi Key: ZSLUVFAKFWKJRC-UHFFFAOYSA-N
SMILES: [Th]
Grade 
  • (2)
  • (1)
Technique 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results