accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7440-30-4

IUPAC Name: thulium
Synonyms: thulium|tulio|thulium atom
Molecular Weight (g/mol): 168.93
Molecular Formula: Tm
InChi Key: FRNOGLGSGLTDKL-UHFFFAOYSA-N
SMILES: [Tm]
Percent Purity 
  • (4)
  • (6)
  • (7)
Quantity 
  • (10)
  • (2)
  • (1)
  • (2)
  • (2)
Form 
  • (3)
  • (2)
  • (2)
Grade 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (10)
  • (2)
  • (7)
  • (4)
  • (6)
  • (7)
  • (10)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)