CAS RN 7440-31-5
Tin mossy, ACS, 99.5% min
CAS: 7440-31-5 Molecular Formula: Sn Molecular Weight (g/mol): 118.71 MDL Number: MFCD00133862 InChI Key: ATJFFYVFTNAWJD-UHFFFAOYSA-N Synonym: powder,stannum,metallic,tin, elemental,wang,zinn,flake,tin, metal,silver matt powder,zinn german PubChem CID: 5352426 ChEBI: CHEBI:32990 IUPAC Name: tin SMILES: [Sn]
Tin foil, 0.05mm (0.002in) thick, 99.999% (metals basis)
CAS: 7440-31-5 Molecular Formula: Sn Molecular Weight (g/mol): 118.71 MDL Number: MFCD00133862 InChI Key: ATJFFYVFTNAWJD-UHFFFAOYSA-N Synonym: powder,stannum,metallic,tin, elemental,wang,zinn,flake,tin, metal,silver matt powder,zinn german PubChem CID: 5352426 ChEBI: CHEBI:32990 IUPAC Name: tin SMILES: [Sn]
Tin powder, -100 mesh, 99.5% (metals basis)
CAS: 7440-31-5 Molecular Formula: Sn Molecular Weight (g/mol): 118.71 MDL Number: MFCD00133862 InChI Key: ATJFFYVFTNAWJD-UHFFFAOYSA-N Synonym: powder,stannum,metallic,tin, elemental,wang,zinn,flake,tin, metal,silver matt powder,zinn german PubChem CID: 5352426 ChEBI: CHEBI:32990 IUPAC Name: tin SMILES: [Sn]
Tin, ACS reagent, 30 mesh
CAS: 7440-31-5 Molecular Formula: Sn Molecular Weight (g/mol): 118.71 MDL Number: MFCD00133862 InChI Key: ATJFFYVFTNAWJD-UHFFFAOYSA-N Synonym: powder,stannum,metallic,tin, elemental,wang,zinn,flake,tin, metal,silver matt powder,zinn german PubChem CID: 5352426 ChEBI: CHEBI:32990 IUPAC Name: tin SMILES: [Sn]
Tin shot, 8-20 mesh, 99.8% (metals basis)
CAS: 7440-31-5 Molecular Formula: Sn Molecular Weight (g/mol): 118.71 MDL Number: MFCD00133862 InChI Key: ATJFFYVFTNAWJD-UHFFFAOYSA-N Synonym: powder,stannum,metallic,tin, elemental,wang,zinn,flake,tin, metal,silver matt powder,zinn german PubChem CID: 5352426 ChEBI: CHEBI:32990 IUPAC Name: tin SMILES: [Sn]
Tin shot, 3.175mm (0.125in) & down, 99% (metals basis)
CAS: 7440-31-5 Molecular Formula: Sn Molecular Weight (g/mol): 118.71 MDL Number: MFCD00133862 InChI Key: ATJFFYVFTNAWJD-UHFFFAOYSA-N Synonym: powder,stannum,metallic,tin, elemental,wang,zinn,flake,tin, metal,silver matt powder,zinn german PubChem CID: 5352426 ChEBI: CHEBI:32990 IUPAC Name: tin SMILES: [Sn]
Tin powder, -100 mesh, 99.995% (metals basis)
CAS: 7440-31-5 Molecular Formula: Sn Molecular Weight (g/mol): 118.71 MDL Number: MFCD00133862 InChI Key: ATJFFYVFTNAWJD-UHFFFAOYSA-N Synonym: powder,stannum,metallic,tin, elemental,wang,zinn,flake,tin, metal,silver matt powder,zinn german PubChem CID: 5352426 ChEBI: CHEBI:32990 IUPAC Name: tin SMILES: [Sn]
Tin, plasma standard solution, Specpure™ Sn 1000μg/mL
CAS: 7440-31-5 Molecular Formula: Sn Molecular Weight (g/mol): 118.71 MDL Number: MFCD00133862 InChI Key: ATJFFYVFTNAWJD-UHFFFAOYSA-N PubChem CID: 5352426 ChEBI: CHEBI:32990 IUPAC Name: tin SMILES: [Sn]
Tin shot, 2-4mm, Puratronic, 99.9999% (metals basis)
CAS: 7440-31-5 Molecular Formula: Sn Molecular Weight (g/mol): 118.71 MDL Number: MFCD00133862 InChI Key: ATJFFYVFTNAWJD-UHFFFAOYSA-N Synonym: powder,stannum,metallic,tin, elemental,wang,zinn,flake,tin, metal,silver matt powder,zinn german PubChem CID: 5352426 ChEBI: CHEBI:32990 IUPAC Name: tin SMILES: [Sn]
Tin wire, 2.0mm (0.08in) dia, Puratronic™, 99.9985% (metals basis)
CAS: 7440-31-5 Molecular Formula: Sn Molecular Weight (g/mol): 118.71 MDL Number: MFCD00133862 InChI Key: ATJFFYVFTNAWJD-UHFFFAOYSA-N Synonym: powder,stannum,metallic,tin, elemental,wang,zinn,flake,tin, metal,silver matt powder,zinn german PubChem CID: 5352426 ChEBI: CHEBI:32990 IUPAC Name: tin SMILES: [Sn]
Tin wire, 0.25mm (0.01in) dia, Puratronic™, 99.998% (metals basis)
CAS: 7440-31-5 Molecular Formula: Sn Molecular Weight (g/mol): 118.71 MDL Number: MFCD00133862 InChI Key: ATJFFYVFTNAWJD-UHFFFAOYSA-N Synonym: powder,stannum,metallic,tin, elemental,wang,zinn,flake,tin, metal,silver matt powder,zinn german PubChem CID: 5352426 ChEBI: CHEBI:32990 IUPAC Name: tin SMILES: [Sn]
Tin slug, 11mm (0.43in) dia x 4mm (0.2in) thick, Puratronic™, 99.999% (metals basis)
CAS: 7440-31-5 Molecular Formula: Sn Molecular Weight (g/mol): 118.71 MDL Number: MFCD00133862 InChI Key: ATJFFYVFTNAWJD-UHFFFAOYSA-N Synonym: powder,stannum,metallic,tin, elemental,wang,zinn,flake,tin, metal,silver matt powder,zinn german PubChem CID: 5352426 ChEBI: CHEBI:32990 IUPAC Name: tin SMILES: [Sn]
Tin rod, 12.7mm (0.5in) dia, 99.98% (metals basis)
CAS: 7440-31-5 Molecular Formula: Sn Molecular Weight (g/mol): 118.71 MDL Number: MFCD00133862 InChI Key: ATJFFYVFTNAWJD-UHFFFAOYSA-N Synonym: powder,stannum,metallic,tin, elemental,wang,zinn,flake,tin, metal,silver matt powder,zinn german PubChem CID: 5352426 ChEBI: CHEBI:32990 IUPAC Name: tin SMILES: [Sn]
Tin, granular, 0.1-0.8 mm
CAS: 7440-31-5 Molecular Formula: Sn Molecular Weight (g/mol): 118.71 MDL Number: MFCD00133862 InChI Key: ATJFFYVFTNAWJD-UHFFFAOYSA-N Synonym: powder,stannum,metallic,tin, elemental,wang,zinn,flake,tin, metal,silver matt powder,zinn german PubChem CID: 5352426 ChEBI: CHEBI:32990 IUPAC Name: tin SMILES: [Sn]
Tin foil, 2.0mm (0.08in) thick, 99.8% (metals basis)
CAS: 7440-31-5 Molecular Formula: Sn Molecular Weight (g/mol): 118.71 MDL Number: MFCD00133862 InChI Key: ATJFFYVFTNAWJD-UHFFFAOYSA-N Synonym: powder,stannum,metallic,tin, elemental,wang,zinn,flake,tin, metal,silver matt powder,zinn german PubChem CID: 5352426 ChEBI: CHEBI:32990 IUPAC Name: tin SMILES: [Sn]