CAS RN 7440-32-6
Titanium sponge, 3-19mm (0.12-0.75in), 99.95% (metals basis)
CAS: 7440-32-6 Molecular Formula: Ti Molecular Weight (g/mol): 47.87 MDL Number: MFCD00011264,MFCD00151301 InChI Key: RTAQQCXQSZGOHL-UHFFFAOYSA-N Synonym: oremet,alloy,cp,vt1,c.p.,contimet 30,jistp28,titanium-125,ii hydride,titan vt 1-1 PubChem CID: 23963 ChEBI: CHEBI:33341 IUPAC Name: titanium SMILES: [Ti]
Titanium foil, 0.25mm (0.01in) thick, annealed, 99.5% (metals basis)
CAS: 7440-32-6 Molecular Formula: Ti Molecular Weight (g/mol): 47.87 MDL Number: MFCD00011264,MFCD00151301 InChI Key: RTAQQCXQSZGOHL-UHFFFAOYSA-N Synonym: oremet,alloy,cp,vt1,c.p.,contimet 30,jistp28,titanium-125,ii hydride,titan vt 1-1 PubChem CID: 23963 ChEBI: CHEBI:33341 IUPAC Name: titanium SMILES: [Ti]
Titanium gauze, 18 mesh woven from 0.28mm (0.011in) dia wire
CAS: 7440-32-6 Molecular Formula: Ti Molecular Weight (g/mol): 47.87 MDL Number: MFCD00011264,MFCD00151301 InChI Key: RTAQQCXQSZGOHL-UHFFFAOYSA-N Synonym: oremet,alloy,cp,vt1,c.p.,contimet 30,jistp28,titanium-125,ii hydride,titan vt 1-1 PubChem CID: 23963 ChEBI: CHEBI:33341 IUPAC Name: titanium SMILES: [Ti]
Titanium wire, 0.5mm (0.02in) dia, 99.98% (metals basis)
CAS: 7440-32-6 Molecular Formula: Ti Molecular Weight (g/mol): 47.87 MDL Number: MFCD00011264,MFCD00151301 InChI Key: RTAQQCXQSZGOHL-UHFFFAOYSA-N Synonym: oremet,alloy,cp,vt1,c.p.,contimet 30,jistp28,titanium-125,ii hydride,titan vt 1-1 PubChem CID: 23963 ChEBI: CHEBI:33341 IUPAC Name: titanium SMILES: [Ti]
Titanium powder, -325 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
CAS: 7440-32-6 Molecular Formula: Ti Molecular Weight (g/mol): 47.87 MDL Number: MFCD00011264,MFCD00151301 InChI Key: RTAQQCXQSZGOHL-UHFFFAOYSA-N Synonym: oremet,alloy,cp,vt1,c.p.,contimet 30,jistp28,titanium-125,ii hydride,titan vt 1-1 PubChem CID: 23963 ChEBI: CHEBI:33341 IUPAC Name: titanium SMILES: [Ti]
Titanium sputtering target, 76.2mm (3.0in) dia x 6.35mm (0.250in) thick, 99.995% (metals basis)
CAS: 7440-32-6 Molecular Formula: Ti Molecular Weight (g/mol): 47.87 MDL Number: MFCD00011264,MFCD00151301 InChI Key: RTAQQCXQSZGOHL-UHFFFAOYSA-N Synonym: oremet,alloy,cp,vt1,c.p.,contimet 30,jistp28,titanium-125,ii hydride,titan vt 1-1 PubChem CID: 23963 ChEBI: CHEBI:33341 IUPAC Name: titanium SMILES: [Ti]
Titanium gauze, 100 mesh woven from 0.05mm (0.002in) dia wire
CAS: 7440-32-6 Molecular Formula: Ti Molecular Weight (g/mol): 47.87 MDL Number: MFCD00011264,MFCD00151301 InChI Key: RTAQQCXQSZGOHL-UHFFFAOYSA-N Synonym: oremet,alloy,cp,vt1,c.p.,contimet 30,jistp28,titanium-125,ii hydride,titan vt 1-1 IUPAC Name: titanium SMILES: [Ti]
Titanium wire, 0.25mm (0.01in) dia, 99.7% (metals basis)
CAS: 7440-32-6 Molecular Formula: Ti Molecular Weight (g/mol): 47.87 MDL Number: MFCD00011264,MFCD00151301 InChI Key: RTAQQCXQSZGOHL-UHFFFAOYSA-N Synonym: oremet,alloy,cp,vt1,c.p.,contimet 30,jistp28,titanium-125,ii hydride,titan vt 1-1 PubChem CID: 23963 ChEBI: CHEBI:33341 IUPAC Name: titanium SMILES: [Ti]
Titanium foil, 0.032mm (0.0013in) thick, annealed, 99.7% (metals basis)
CAS: 7440-32-6 Molecular Formula: Ti Molecular Weight (g/mol): 47.87 MDL Number: MFCD00011264,MFCD00151301 InChI Key: RTAQQCXQSZGOHL-UHFFFAOYSA-N Synonym: oremet,alloy,cp,vt1,c.p.,contimet 30,jistp28,titanium-125,ii hydride,titan vt 1-1 PubChem CID: 23963 ChEBI: CHEBI:33341 IUPAC Name: titanium SMILES: [Ti]
Titanium slug, 9.5mm (0.37in) dia x 9.5mm (0.37in) length, 99.995% (metals basis)
CAS: 7440-32-6 Molecular Formula: Ti Molecular Weight (g/mol): 47.87 MDL Number: MFCD00011264,MFCD00151301 InChI Key: RTAQQCXQSZGOHL-UHFFFAOYSA-N Synonym: oremet,alloy,cp,vt1,c.p.,contimet 30,jistp28,titanium-125,ii hydride,titan vt 1-1 PubChem CID: 23963 ChEBI: CHEBI:33341 IUPAC Name: titanium SMILES: [Ti]
Titanium plate, 3.175mm (0.125in) thick, 100x100mm (3.9x3.9in), 99.99% (metals basis)
CAS: 7440-32-6 Molecular Formula: Ti Molecular Weight (g/mol): 47.87 MDL Number: MFCD00011264,MFCD00151301 InChI Key: RTAQQCXQSZGOHL-UHFFFAOYSA-N Synonym: oremet,alloy,cp,vt1,c.p.,contimet 30,jistp28,titanium-125,ii hydride,titan vt 1-1 PubChem CID: 23963 ChEBI: CHEBI:33341 IUPAC Name: titanium SMILES: [Ti]
Titanium gauze, 30 mesh woven from 0.102mm (0.004in) dia wire
CAS: 7440-32-6 Molecular Formula: Ti Molecular Weight (g/mol): 47.87 MDL Number: MFCD00011264,MFCD00151301 InChI Key: RTAQQCXQSZGOHL-UHFFFAOYSA-N Synonym: oremet,alloy,cp,vt1,c.p.,contimet 30,jistp28,titanium-125,ii hydride,titan vt 1-1 PubChem CID: 23963 ChEBI: CHEBI:33341 IUPAC Name: titanium SMILES: [Ti]
Titanium, plasma standard solution, Specpure™ Ti 10,000μg/mL
CAS: 7440-32-6 Molecular Formula: Ti Molecular Weight (g/mol): 47.87 MDL Number: MFCD00011264,MFCD00151301 InChI Key: RTAQQCXQSZGOHL-UHFFFAOYSA-N Synonym: oremet,alloy,cp,vt1,c.p.,contimet 30,jistp28,titanium-125,ii hydride,titan vt 1-1 PubChem CID: 23963 ChEBI: CHEBI:33341 IUPAC Name: titanium SMILES: [Ti]
Titanium gauze, 20 mesh woven from 0.076mm (0.003in) dia wire
CAS: 7440-32-6 Molecular Formula: Ti Molecular Weight (g/mol): 47.87 MDL Number: MFCD00011264,MFCD00151301 InChI Key: RTAQQCXQSZGOHL-UHFFFAOYSA-N Synonym: oremet,alloy,cp,vt1,c.p.,contimet 30,jistp28,titanium-125,ii hydride,titan vt 1-1 PubChem CID: 23963 ChEBI: CHEBI:33341 IUPAC Name: titanium SMILES: [Ti]
Titanium wire, 1.0mm (0.04in) dia, hard, 99.99% (metals basis)
CAS: 7440-32-6 Molecular Formula: Ti Molecular Weight (g/mol): 47.87 MDL Number: MFCD00011264,MFCD00151301 InChI Key: RTAQQCXQSZGOHL-UHFFFAOYSA-N Synonym: oremet,alloy,cp,vt1,c.p.,contimet 30,jistp28,titanium-125,ii hydride,titan vt 1-1 PubChem CID: 23963 ChEBI: CHEBI:33341 IUPAC Name: titanium SMILES: [Ti]