CAS RN 7440-45-1
Cerium powder, -60 mesh, 99.9% (REO)
CAS: 7440-45-1 Molecular Formula: Ce Molecular Weight (g/mol): 140.12 MDL Number: MFCD00010924 InChI Key: GWXLDORMOJMVQZ-UHFFFAOYSA-N Synonym: cerio,ingot,cerium, chips,foil, 3n,zer,ceramics-aeium,acmc-1bl5r,chips, 3n reo,powder,-200 mesh,cerium PubChem CID: 23974 ChEBI: CHEBI:33369 IUPAC Name: cerium SMILES: [Ce]
Cerium foil, 0.25mm (0.01in) thick, 99.9% (REO)
CAS: 7440-45-1 Molecular Formula: Ce Molecular Weight (g/mol): 140.12 MDL Number: MFCD00010924 InChI Key: GWXLDORMOJMVQZ-UHFFFAOYSA-N Synonym: cerio,ingot,cerium, chips,foil, 3n,zer,ceramics-aeium,acmc-1bl5r,chips, 3n reo,powder,-200 mesh,cerium PubChem CID: 23974 ChEBI: CHEBI:33369 IUPAC Name: cerium SMILES: [Ce]
Cerium foil, 0.62mm (0.024in) thick, 99.9% (REO)
CAS: 7440-45-1 Molecular Formula: Ce Molecular Weight (g/mol): 140.12 MDL Number: MFCD00010924 InChI Key: GWXLDORMOJMVQZ-UHFFFAOYSA-N Synonym: cerio,ingot,cerium, chips,foil, 3n,zer,ceramics-aeium,acmc-1bl5r,chips, 3n reo,powder,-200 mesh,cerium PubChem CID: 23974 ChEBI: CHEBI:33369 IUPAC Name: cerium SMILES: [Ce]
Cerium, plasma standard solution, Specpure™ Ce 1000μg/mL
CAS: 7440-45-1 Molecular Formula: Ce Molecular Weight (g/mol): 140.12 MDL Number: MFCD00010924 InChI Key: GWXLDORMOJMVQZ-UHFFFAOYSA-N PubChem CID: 23974 ChEBI: CHEBI:33369 IUPAC Name: cerium SMILES: [Ce]
Cerium rod, 6.35mm (0.25in) dia, 99.9% (metals basis excluding Ta)
CAS: 7440-45-1 Molecular Formula: Ce Molecular Weight (g/mol): 140.12 MDL Number: MFCD00010924 InChI Key: GWXLDORMOJMVQZ-UHFFFAOYSA-N Synonym: cerio,ingot,cerium, chips,foil, 3n,zer,ceramics-aeium,acmc-1bl5r,chips, 3n reo,powder,-200 mesh,cerium PubChem CID: 23974 ChEBI: CHEBI:33369 IUPAC Name: cerium SMILES: [Ce]
Cerium foil, 1.0mm (0.04in) thick, 99.9% (REO)
CAS: 7440-45-1 Molecular Formula: Ce Molecular Weight (g/mol): 140.12 MDL Number: MFCD00010924 InChI Key: GWXLDORMOJMVQZ-UHFFFAOYSA-N Synonym: cerio,ingot,cerium, chips,foil, 3n,zer,ceramics-aeium,acmc-1bl5r,chips, 3n reo,powder,-200 mesh,cerium PubChem CID: 23974 ChEBI: CHEBI:33369 IUPAC Name: cerium SMILES: [Ce]
Cerium foil, 0.1mm (0.004in) thick, 99.9% (REO)
CAS: 7440-45-1 Molecular Formula: Ce Molecular Weight (g/mol): 140.12 MDL Number: MFCD00010924 InChI Key: GWXLDORMOJMVQZ-UHFFFAOYSA-N Synonym: cerio,ingot,cerium, chips,foil, 3n,zer,ceramics-aeium,acmc-1bl5r,chips, 3n reo,powder,-200 mesh,cerium PubChem CID: 23974 ChEBI: CHEBI:33369 IUPAC Name: cerium SMILES: [Ce]
Cerium ingot, 99.8% min (REO)
CAS: 7440-45-1 Molecular Formula: Ce Molecular Weight (g/mol): 140.12 MDL Number: MFCD00010924 InChI Key: GWXLDORMOJMVQZ-UHFFFAOYSA-N Synonym: cerio,ingot,cerium, chips,foil, 3n,zer,ceramics-aeium,acmc-1bl5r,chips, 3n reo,powder,-200 mesh,cerium PubChem CID: 23974 ChEBI: CHEBI:33369 IUPAC Name: cerium SMILES: [Ce]
Cerium rod, 12.7mm (0.5in) dia, 99.8% (metals basis excluding Ta), Thermo Scientific Chemicals
CAS: 7440-45-1 Molecular Formula: Ce Molecular Weight (g/mol): 140.12 MDL Number: MFCD00010924 InChI Key: GWXLDORMOJMVQZ-UHFFFAOYSA-N Synonym: cerio,ingot,cerium, chips,foil, 3n,zer,ceramics-aeium,acmc-1bl5r,chips, 3n reo,powder,-200 mesh,cerium PubChem CID: 23974 ChEBI: CHEBI:33369 IUPAC Name: cerium SMILES: [Ce]
Cerium ingot, 99% (REO)
CAS: 7440-45-1 Molecular Formula: Ce Molecular Weight (g/mol): 140.12 MDL Number: MFCD00010924 InChI Key: GWXLDORMOJMVQZ-UHFFFAOYSA-N Synonym: cerio,ingot,cerium, chips,foil, 3n,zer,ceramics-aeium,acmc-1bl5r,chips, 3n reo,powder,-200 mesh,cerium PubChem CID: 23974 ChEBI: CHEBI:33369 IUPAC Name: cerium SMILES: [Ce]
Cerium sputtering target, 76.2mm dia x 3.18mm thick
CAS: 7440-45-1 Molecular Formula: Ce Molecular Weight (g/mol): 140.12 MDL Number: MFCD00010924 InChI Key: GWXLDORMOJMVQZ-UHFFFAOYSA-N Synonym: cerio,ingot,cerium, chips,foil, 3n,zer,ceramics-aeium,acmc-1bl5r,chips, 3n reo,powder,-200 mesh,cerium PubChem CID: 23974 ChEBI: CHEBI:33369 IUPAC Name: cerium SMILES: [Ce]
Cerium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™
CAS: 7440-45-1 Molecular Formula: Ce Molecular Weight (g/mol): 140.12 MDL Number: MFCD00010924 InChI Key: GWXLDORMOJMVQZ-UHFFFAOYSA-N PubChem CID: 23974 ChEBI: CHEBI:33369 IUPAC Name: cerium SMILES: [Ce]
Cerium Standard For ICP, 10,000μg/mL in 5% HNO3, SPEX CertiPrep™
CAS: 7440-45-1 Molecular Formula: Ce Molecular Weight (g/mol): 140.12 MDL Number: MFCD00010924 InChI Key: GWXLDORMOJMVQZ-UHFFFAOYSA-N PubChem CID: 23974 ChEBI: CHEBI:33369 IUPAC Name: cerium SMILES: [Ce]
Sigma Aldrich Cerium
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Cerium
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
- 1
- 2