CAS RN 7440-47-3
Chromium powder, -325 mesh, 99% (metals basis)
CAS: 7440-47-3 Molecular Formula: Cr Molecular Weight (g/mol): 52.00 MDL Number: MFCD00010944 InChI Key: VYZAMTAEIAYCRO-UHFFFAOYSA-N Synonym: chrom,chrome,cromo,metal,chrome french,compounds,chrom german,chromium, elemental,chromium, metal,ccris 159 PubChem CID: 23976 ChEBI: CHEBI:28073 IUPAC Name: chromium SMILES: [Cr]
Chromium powder, -60 mesh, 99.95% (metals basis)
CAS: 7440-47-3 Molecular Formula: Cr Molecular Weight (g/mol): 52.00 MDL Number: MFCD00010944 InChI Key: VYZAMTAEIAYCRO-UHFFFAOYSA-N Synonym: chrom,chrome,cromo,metal,chrome french,compounds,chrom german,chromium, elemental,chromium, metal,ccris 159 PubChem CID: 23976 ChEBI: CHEBI:28073 IUPAC Name: chromium SMILES: [Cr]
Chromium powder, -100 mesh, 99% (metals basis)
CAS: 7440-47-3 Molecular Formula: Cr Molecular Weight (g/mol): 52.00 MDL Number: MFCD00010944 InChI Key: VYZAMTAEIAYCRO-UHFFFAOYSA-N Synonym: chrom,chrome,cromo,metal,chrome french,compounds,chrom german,chromium, elemental,chromium, metal,ccris 159 PubChem CID: 23976 ChEBI: CHEBI:28073 IUPAC Name: chromium SMILES: [Cr]
Chromium pieces, electrolytic, 6mm & down, Puratronic™, 99.997% (metals basis)
CAS: 7440-47-3 Molecular Formula: Cr Molecular Weight (g/mol): 52.00 MDL Number: MFCD00010944 InChI Key: VYZAMTAEIAYCRO-UHFFFAOYSA-N Synonym: chrom,chrome,cromo,metal,chrome french,compounds,chrom german,chromium, elemental,chromium, metal,ccris 159 PubChem CID: 23976 ChEBI: CHEBI:28073 IUPAC Name: chromium SMILES: [Cr]
Chromium pieces, 1-25mm (0.04-1in), 99.99% (metals basis)
CAS: 7440-47-3 Molecular Formula: Cr Molecular Weight (g/mol): 52.00 MDL Number: MFCD00010944 InChI Key: VYZAMTAEIAYCRO-UHFFFAOYSA-N Synonym: chrom,chrome,cromo,metal,chrome french,compounds,chrom german,chromium, elemental,chromium, metal,ccris 159 PubChem CID: 23976 ChEBI: CHEBI:28073 IUPAC Name: chromium SMILES: [Cr]
Chromium powder, -100+325 mesh, 99.97% (metals basis)
CAS: 7440-47-3 Molecular Formula: Cr Molecular Weight (g/mol): 52.00 MDL Number: MFCD00010944 InChI Key: VYZAMTAEIAYCRO-UHFFFAOYSA-N Synonym: chrom,chrome,cromo,metal,chrome french,compounds,chrom german,chromium, elemental,chromium, metal,ccris 159 PubChem CID: 23976 ChEBI: CHEBI:28073 IUPAC Name: chromium SMILES: [Cr]
Chromium chunks, 2-15mm (0.08-0.59in), 99.996% (metals basis)
CAS: 7440-47-3 Molecular Formula: Cr Molecular Weight (g/mol): 52.00 MDL Number: MFCD00010944 InChI Key: VYZAMTAEIAYCRO-UHFFFAOYSA-N Synonym: chrom,chrome,cromo,metal,chrome french,compounds,chrom german,chromium, elemental,chromium, metal,ccris 159 PubChem CID: 23976 ChEBI: CHEBI:28073 IUPAC Name: chromium SMILES: [Cr]
Chromium pieces, 2-8mm (0.08-0.31in), 99.99% (metals basis)
CAS: 7440-47-3 Molecular Formula: Cr Molecular Weight (g/mol): 52.00 MDL Number: MFCD00010944 InChI Key: VYZAMTAEIAYCRO-UHFFFAOYSA-N Synonym: chrom,chrome,cromo,metal,chrome french,compounds,chrom german,chromium, elemental,chromium, metal,ccris 159 PubChem CID: 23976 ChEBI: CHEBI:28073 IUPAC Name: chromium SMILES: [Cr]
Chromium crystallite pieces, 2-15mm (0.08-0.59in), 99.996% (metals basis)
CAS: 7440-47-3 Molecular Formula: Cr Molecular Weight (g/mol): 52.00 MDL Number: MFCD00010944 InChI Key: VYZAMTAEIAYCRO-UHFFFAOYSA-N Synonym: chrom,chrome,cromo,metal,chrome french,compounds,chrom german,chromium, elemental,chromium, metal,ccris 159 PubChem CID: 23976 ChEBI: CHEBI:28073 IUPAC Name: chromium SMILES: [Cr]
Chromium powder, APS <10 micron, 99.2% (metals basis)
CAS: 7440-47-3 Molecular Formula: Cr Molecular Weight (g/mol): 52.00 MDL Number: MFCD00010944 InChI Key: VYZAMTAEIAYCRO-UHFFFAOYSA-N Synonym: chrom,chrome,cromo,metal,chrome french,compounds,chrom german,chromium, elemental,chromium, metal,ccris 159 PubChem CID: 23976 ChEBI: CHEBI:28073 IUPAC Name: chromium SMILES: [Cr]
Chromium granules, -4+7 mesh, 99.999% (metals basis)
CAS: 7440-47-3 Molecular Formula: Cr Molecular Weight (g/mol): 52.00 MDL Number: MFCD00010944 InChI Key: VYZAMTAEIAYCRO-UHFFFAOYSA-N Synonym: chrom,chrome,cromo,metal,chrome french,compounds,chrom german,chromium, elemental,chromium, metal,ccris 159 PubChem CID: 23976 ChEBI: CHEBI:28073 IUPAC Name: chromium SMILES: [Cr]
Chromium sputtering target, 50.8mm (2.0in) dia x 3.18mm (0.125in) thick, 99.95% (metals basis)
CAS: 7440-47-3 Molecular Formula: Cr Molecular Weight (g/mol): 52.00 MDL Number: MFCD00010944 InChI Key: VYZAMTAEIAYCRO-UHFFFAOYSA-N Synonym: chrom,chrome,cromo,metal,chrome french,compounds,chrom german,chromium, elemental,chromium, metal,ccris 159 PubChem CID: 23976 ChEBI: CHEBI:28073 IUPAC Name: chromium SMILES: [Cr]
Chromium pieces, 2-3mm (0.08-0.12in), 99.995% (metals basis)
CAS: 7440-47-3 Molecular Formula: Cr Molecular Weight (g/mol): 52.00 MDL Number: MFCD00010944 InChI Key: VYZAMTAEIAYCRO-UHFFFAOYSA-N Synonym: chrom,chrome,cromo,metal,chrome french,compounds,chrom german,chromium, elemental,chromium, metal,ccris 159 PubChem CID: 23976 ChEBI: CHEBI:28073 IUPAC Name: chromium SMILES: [Cr]
Chromium, 99+%, pure, powder
CAS: 7440-47-3 Molecular Formula: Cr Molecular Weight (g/mol): 52.00 MDL Number: MFCD00010944 InChI Key: VYZAMTAEIAYCRO-UHFFFAOYSA-N Synonym: chrom,chrome,cromo,metal,chrome french,compounds,chrom german,chromium, elemental,chromium, metal,ccris 159 PubChem CID: 23976 ChEBI: CHEBI:28073 IUPAC Name: chromium SMILES: [Cr]
Chromium, plasma standard solution, Specpure™ Cr 1000μg/mL
CAS: 7440-47-3 Molecular Formula: Cr Molecular Weight (g/mol): 52.00 MDL Number: MFCD00010944 InChI Key: VYZAMTAEIAYCRO-UHFFFAOYSA-N PubChem CID: 23976 ChEBI: CHEBI:28073 IUPAC Name: chromium SMILES: [Cr]