CAS RN 7440-50-8
Copper AA Standard, 1000 ppm Cu in 3% HNO3, Ricca Chemical
CAS: 7440-50-8 Molecular Formula: Cu Molecular Weight (g/mol): 63.55 MDL Number: MFCD00010965 InChI Key: RYGMFSIKBFXOCR-UHFFFAOYSA-N PubChem CID: 23978 ChEBI: CHEBI:30052 IUPAC Name: copper SMILES: [Cu]
Copper wire, 1.0mm (0.04in) dia, annealed, 99.9% (metals basis)
CAS: 7440-50-8 Molecular Formula: Cu Molecular Weight (g/mol): 63.55 MDL Number: MFCD00010965 InChI Key: RYGMFSIKBFXOCR-UHFFFAOYSA-N Synonym: cuprum,cobre,cuivre,blister,cathode,bronze,powder,anode,precipitates,kupfer PubChem CID: 23978 ChEBI: CHEBI:30052 IUPAC Name: copper SMILES: [Cu]
Copper foil, 0.1mm (0.004in) thick, Puratronic™, 99.999% (metals basis)
CAS: 7440-50-8 Molecular Formula: Cu Molecular Weight (g/mol): 63.55 MDL Number: MFCD00010965 InChI Key: RYGMFSIKBFXOCR-UHFFFAOYSA-N Synonym: cuprum,cobre,cuivre,blister,cathode,bronze,powder,anode,precipitates,kupfer PubChem CID: 23978 ChEBI: CHEBI:30052 IUPAC Name: copper SMILES: [Cu]
Copper shot, 0.8-2mm (0.03-0.08in), 99.5% (metals basis)
CAS: 7440-50-8 Molecular Formula: Cu MDL Number: MFCD00010965 InChI Key: RYGMFSIKBFXOCR-UHFFFAOYSA-N Synonym: cuprum,cobre,cuivre,blister,cathode,bronze,powder,anode,precipitates,kupfer PubChem CID: 23978 ChEBI: CHEBI:30052 IUPAC Name: copper SMILES: [Cu]
Copper powder, spherical, APS 10 micron, 99.9% (metals basis)
CAS: 7440-50-8 Molecular Formula: Cu Molecular Weight (g/mol): 63.55 MDL Number: MFCD00010965 InChI Key: RYGMFSIKBFXOCR-UHFFFAOYSA-N Synonym: cuprum,cobre,cuivre,blister,cathode,bronze,powder,anode,precipitates,kupfer PubChem CID: 23978 ChEBI: CHEBI:30052 IUPAC Name: copper SMILES: [Cu]
Copper powder, -625 mesh, APS 3.25-4.75 micron, 99.9% (metals basis)
CAS: 7440-50-8 Molecular Formula: Cu Molecular Weight (g/mol): 63.55 MDL Number: MFCD00010965 InChI Key: RYGMFSIKBFXOCR-UHFFFAOYSA-N Synonym: cuprum,cobre,cuivre,blister,cathode,bronze,powder,anode,precipitates,kupfer PubChem CID: 23978 ChEBI: CHEBI:30052 IUPAC Name: copper SMILES: [Cu]
Copper, 99.9%, (trace metal basis), shot 1-10 mm
CAS: 7440-50-8 Molecular Formula: Cu Molecular Weight (g/mol): 63.55 MDL Number: MFCD00010965 InChI Key: RYGMFSIKBFXOCR-UHFFFAOYSA-N Synonym: cuprum,cobre,cuivre,blister,cathode,bronze,powder,anode,precipitates,kupfer PubChem CID: 23978 ChEBI: CHEBI:30052 IUPAC Name: copper SMILES: [Cu]
Copper shot, 1-10mm (0.04-0.4in), 99.9% (metals basis)
CAS: 7440-50-8 Molecular Formula: Cu Molecular Weight (g/mol): 63.55 MDL Number: MFCD00010965 InChI Key: RYGMFSIKBFXOCR-UHFFFAOYSA-N Synonym: cuprum,cobre,cuivre,blister,cathode,bronze,powder,anode,precipitates,kupfer PubChem CID: 23978 ChEBI: CHEBI:30052 IUPAC Name: copper SMILES: [Cu]
Copper foil, 0.025mm (0.001in) thick, annealed, uncoated, 99.8% (metals basis)
CAS: 7440-50-8 Molecular Formula: Cu Molecular Weight (g/mol): 63.55 MDL Number: MFCD00010965 InChI Key: RYGMFSIKBFXOCR-UHFFFAOYSA-N Synonym: cuprum,cobre,cuivre,blister,cathode,bronze,powder,anode,precipitates,kupfer PubChem CID: 23978 ChEBI: CHEBI:30052 IUPAC Name: copper SMILES: [Cu]
Copper foil, 0.675mm (0.027in) thick, annealed, 99.9% (metals basis)
CAS: 7440-50-8 Molecular Formula: Cu Molecular Weight (g/mol): 63.55 MDL Number: MFCD00010965 InChI Key: RYGMFSIKBFXOCR-UHFFFAOYSA-N Synonym: cuprum,cobre,cuivre,blister,cathode,bronze,powder,anode,precipitates,kupfer PubChem CID: 23978 ChEBI: CHEBI:30052 IUPAC Name: copper SMILES: [Cu]
Copper wire, 1.5mm (0.06in) dia, Puratronic™, 99.9999% (metals basis)
CAS: 7440-50-8 Molecular Formula: Cu Molecular Weight (g/mol): 63.55 MDL Number: MFCD00010965 InChI Key: RYGMFSIKBFXOCR-UHFFFAOYSA-N Synonym: cuprum,cobre,cuivre,blister,cathode,bronze,powder,anode,precipitates,kupfer PubChem CID: 23978 ChEBI: CHEBI:30052 IUPAC Name: copper SMILES: [Cu]
Copper powder, spherical, -170+270 mesh, 99.9% (metals basis)
CAS: 7440-50-8 Molecular Formula: Cu Molecular Weight (g/mol): 63.55 MDL Number: MFCD00010965 InChI Key: RYGMFSIKBFXOCR-UHFFFAOYSA-N Synonym: cuprum,cobre,cuivre,blister,cathode,bronze,powder,anode,precipitates,kupfer PubChem CID: 23978 ChEBI: CHEBI:30052 IUPAC Name: copper SMILES: [Cu]
Copper powder, -325 mesh, 10% max +325 mesh, 99% (metals basis)
CAS: 7440-50-8 Molecular Formula: Cu Molecular Weight (g/mol): 63.55 MDL Number: MFCD00010965 InChI Key: RYGMFSIKBFXOCR-UHFFFAOYSA-N Synonym: cuprum,cobre,cuivre,blister,cathode,bronze,powder,anode,precipitates,kupfer PubChem CID: 23978 ChEBI: CHEBI:30052 IUPAC Name: copper SMILES: [Cu]
Copper wire, 0.25mm (0.01in) dia, Puratronic™, 99.9999% (metals basis)
CAS: 7440-50-8 Molecular Formula: Cu Molecular Weight (g/mol): 63.55 MDL Number: MFCD00010965 InChI Key: RYGMFSIKBFXOCR-UHFFFAOYSA-N Synonym: cuprum,cobre,cuivre,blister,cathode,bronze,powder,anode,precipitates,kupfer PubChem CID: 23978 ChEBI: CHEBI:30052 IUPAC Name: copper SMILES: [Cu]
Copper slug, 6.35mm (0.25 in.) dia. x 6.35mm (0.25 in.) length, Puratronic™, 99.9999% (metals basis), Thermo Scientific Chemicals
CAS: 7440-50-8 Molecular Formula: Cu Molecular Weight (g/mol): 63.55 MDL Number: MFCD00010965 InChI Key: RYGMFSIKBFXOCR-UHFFFAOYSA-N Synonym: cuprum,cobre,cuivre,blister,cathode,bronze,powder,anode,precipitates,kupfer PubChem CID: 23978 ChEBI: CHEBI:30052 IUPAC Name: copper SMILES: [Cu]