CAS RN 7440-52-0
Erbium foil, 0.25mm (0.01in) thick, 99.9% (REO)
CAS: 7440-52-0 Molecular Formula: Er Molecular Weight (g/mol): 167.26 MDL Number: MFCD00010987 InChI Key: UYAHIZSMUZPPFV-UHFFFAOYSA-N Synonym: unii-77b218d3ye,foil,iii ion,erbio,pieces,powder,chips,ingot,er3,pieces, distilled dendritic PubChem CID: 23980 ChEBI: CHEBI:33379 IUPAC Name: erbium SMILES: [Er]
Erbium powder, -40 mesh, 99.9% (REO)
CAS: 7440-52-0 Molecular Formula: Er Molecular Weight (g/mol): 167.26 MDL Number: MFCD00010987 InChI Key: UYAHIZSMUZPPFV-UHFFFAOYSA-N Synonym: unii-77b218d3ye,foil,iii ion,erbio,pieces,powder,chips,ingot,er3,pieces, distilled dendritic PubChem CID: 23980 ChEBI: CHEBI:33379 IUPAC Name: erbium SMILES: [Er]
Erbium rod, 12.7mm (0.5in) dia, 99.9% (metals basis excluding Ta)
CAS: 7440-52-0 Molecular Formula: Er Molecular Weight (g/mol): 167.26 MDL Number: MFCD00010987 InChI Key: UYAHIZSMUZPPFV-UHFFFAOYSA-N Synonym: unii-77b218d3ye,foil,iii ion,erbio,pieces,powder,chips,ingot,er3,pieces, distilled dendritic PubChem CID: 23980 ChEBI: CHEBI:33379 IUPAC Name: erbium SMILES: [Er]
Erbium foil, 0.62mm (0.024in) thick, 99.9% (REO)
CAS: 7440-52-0 Molecular Formula: Er Molecular Weight (g/mol): 167.26 MDL Number: MFCD00010987 InChI Key: UYAHIZSMUZPPFV-UHFFFAOYSA-N Synonym: unii-77b218d3ye,foil,iii ion,erbio,pieces,powder,chips,ingot,er3,pieces, distilled dendritic PubChem CID: 23980 ChEBI: CHEBI:33379 IUPAC Name: erbium SMILES: [Er]
Erbium rod, 6.35mm (0.25in) dia, 99.9% (metals basis excluding Ta)
CAS: 7440-52-0 Molecular Formula: Er Molecular Weight (g/mol): 167.26 MDL Number: MFCD00010987 InChI Key: UYAHIZSMUZPPFV-UHFFFAOYSA-N Synonym: unii-77b218d3ye,foil,iii ion,erbio,pieces,powder,chips,ingot,er3,pieces, distilled dendritic PubChem CID: 23980 ChEBI: CHEBI:33379 IUPAC Name: erbium SMILES: [Er]
Erbium pieces, distilled dendritic, 99.9% (REO), Thermo Scientific Chemicals
CAS: 7440-52-0 Molecular Formula: Er Molecular Weight (g/mol): 167.26 MDL Number: MFCD00010987 InChI Key: UYAHIZSMUZPPFV-UHFFFAOYSA-N Synonym: unii-77b218d3ye,foil,iii ion,erbio,pieces,powder,chips,ingot,er3,pieces, distilled dendritic PubChem CID: 23980 ChEBI: CHEBI:33379 IUPAC Name: erbium SMILES: [Er]
Erbium foil, 0.1mm (0.004in) thick, 99.9% (REO), Thermo Scientific Chemicals
CAS: 7440-52-0 Molecular Formula: Er Molecular Weight (g/mol): 167.26 MDL Number: MFCD00010987 InChI Key: UYAHIZSMUZPPFV-UHFFFAOYSA-N Synonym: unii-77b218d3ye,foil,iii ion,erbio,pieces,powder,chips,ingot,er3,pieces, distilled dendritic PubChem CID: 23980 ChEBI: CHEBI:33379 IUPAC Name: erbium SMILES: [Er]
Erbium foil, 0.025mm (0.001 in.) thick, 99.9% (REO), Thermo Scientific Chemicals
CAS: 7440-52-0 Molecular Formula: Er Molecular Weight (g/mol): 167.26 MDL Number: MFCD00010987 InChI Key: UYAHIZSMUZPPFV-UHFFFAOYSA-N Synonym: unii-77b218d3ye,foil,iii ion,erbio,pieces,powder,chips,ingot,er3,pieces, distilled dendritic PubChem CID: 23980 ChEBI: CHEBI:33379 IUPAC Name: erbium SMILES: [Er]
Erbium pieces, 99.9% (REO)
CAS: 7440-52-0 Molecular Formula: Er Molecular Weight (g/mol): 167.26 MDL Number: MFCD00010987 InChI Key: UYAHIZSMUZPPFV-UHFFFAOYSA-N Synonym: unii-77b218d3ye,foil,iii ion,erbio,pieces,powder,chips,ingot,er3,pieces, distilled dendritic PubChem CID: 23980 ChEBI: CHEBI:33379 IUPAC Name: erbium SMILES: [Er]
Sigma Aldrich Erbium
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Erbium
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Erbium
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Erbium
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Erbium
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Erbium
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
- 1
- 2