CAS RN 7440-54-2
Gadolinium powder, -40 mesh, 99% (REO)
CAS: 7440-54-2 Molecular Formula: Gd Molecular Weight (g/mol): 157.25 MDL Number: MFCD00011022 InChI Key: UIWYJDYFSGRHKR-UHFFFAOYSA-N Synonym: atom,gadolinio,unii-au0v1lm3jt,trihydride,au0v1lm3jt,foil, 3n,gadolinium, elemental,ingot,gadolinium, chips,compounds PubChem CID: 23982 ChEBI: CHEBI:33375 IUPAC Name: gadolinium SMILES: [Gd]
Gadolinium rod, 12.7mm (0.5in) dia, 99.9% (metals basis excluding Ta)
CAS: 7440-54-2 Molecular Formula: Gd Molecular Weight (g/mol): 157.25 MDL Number: MFCD00011022 InChI Key: UIWYJDYFSGRHKR-UHFFFAOYSA-N Synonym: atom,gadolinio,unii-au0v1lm3jt,trihydride,au0v1lm3jt,foil, 3n,gadolinium, elemental,ingot,gadolinium, chips,compounds PubChem CID: 23982 ChEBI: CHEBI:33375 IUPAC Name: gadolinium SMILES: [Gd]
Gadolinium ingot/button, =50.8mm (2.0in) dia, 99% (REO)
CAS: 7440-54-2 Molecular Formula: Gd Molecular Weight (g/mol): 157.25 MDL Number: MFCD00011022 InChI Key: UIWYJDYFSGRHKR-UHFFFAOYSA-N Synonym: atom,gadolinio,unii-au0v1lm3jt,trihydride,au0v1lm3jt,foil, 3n,gadolinium, elemental,ingot,gadolinium, chips,compounds PubChem CID: 23982 ChEBI: CHEBI:33375 IUPAC Name: gadolinium SMILES: [Gd]
Gadolinium powder, -200 mesh, 99.9% (REO)
CAS: 7440-54-2 Molecular Formula: Gd Molecular Weight (g/mol): 157.25 MDL Number: MFCD00011022 InChI Key: UIWYJDYFSGRHKR-UHFFFAOYSA-N Synonym: atom,gadolinio,unii-au0v1lm3jt,trihydride,au0v1lm3jt,foil, 3n,gadolinium, elemental,ingot,gadolinium, chips,compounds PubChem CID: 23982 ChEBI: CHEBI:33375 IUPAC Name: gadolinium SMILES: [Gd]
Gadolinium rod, 6.35mm (0.25in) dia, 99.9% (metals basis excluding Ta)
CAS: 7440-54-2 Molecular Formula: Gd Molecular Weight (g/mol): 157.25 MDL Number: MFCD00011022 InChI Key: UIWYJDYFSGRHKR-UHFFFAOYSA-N Synonym: atom,gadolinio,unii-au0v1lm3jt,trihydride,au0v1lm3jt,foil, 3n,gadolinium, elemental,ingot,gadolinium, chips,compounds PubChem CID: 23982 ChEBI: CHEBI:33375 IUPAC Name: gadolinium SMILES: [Gd]
Gadolinium foil, 0.25mm (0.01in) thick, 99.9% (REO)
CAS: 7440-54-2 Molecular Formula: Gd Molecular Weight (g/mol): 157.25 MDL Number: MFCD00011022 InChI Key: UIWYJDYFSGRHKR-UHFFFAOYSA-N Synonym: atom,gadolinio,unii-au0v1lm3jt,trihydride,au0v1lm3jt,foil, 3n,gadolinium, elemental,ingot,gadolinium, chips,compounds PubChem CID: 23982 ChEBI: CHEBI:33375 IUPAC Name: gadolinium SMILES: [Gd]
Gadolinium pieces, 20mm (0.79in) & down, 99.9% (REO)
CAS: 7440-54-2 Molecular Formula: Gd Molecular Weight (g/mol): 157.25 MDL Number: MFCD00011022 InChI Key: UIWYJDYFSGRHKR-UHFFFAOYSA-N Synonym: atom,gadolinio,unii-au0v1lm3jt,trihydride,au0v1lm3jt,foil, 3n,gadolinium, elemental,ingot,gadolinium, chips,compounds PubChem CID: 23982 ChEBI: CHEBI:33375 IUPAC Name: gadolinium SMILES: [Gd]
Gadolinium foil, 0.1mm (0.004in) thick, 99.9% (REO)
CAS: 7440-54-2 Molecular Formula: Gd Molecular Weight (g/mol): 157.25 MDL Number: MFCD00011022 InChI Key: UIWYJDYFSGRHKR-UHFFFAOYSA-N Synonym: atom,gadolinio,unii-au0v1lm3jt,trihydride,au0v1lm3jt,foil, 3n,gadolinium, elemental,ingot,gadolinium, chips,compounds PubChem CID: 23982 ChEBI: CHEBI:33375 IUPAC Name: gadolinium SMILES: [Gd]
Gadolinium rod, 19mm (0.75 in.) dia., 99.9% (metals basis excluding Ta), Thermo Scientific™
CAS: 7440-54-2 Molecular Formula: Gd Molecular Weight (g/mol): 157.25 MDL Number: MFCD00011022 InChI Key: UIWYJDYFSGRHKR-UHFFFAOYSA-N Synonym: atom,gadolinio,unii-au0v1lm3jt,trihydride,au0v1lm3jt,foil, 3n,gadolinium, elemental,ingot,gadolinium, chips,compounds PubChem CID: 23982 ChEBI: CHEBI:33375 IUPAC Name: gadolinium SMILES: [Gd]
Gadolinium sputtering target, 50.8mm dia x 3.18mm thick
CAS: 7440-54-2 Molecular Formula: Gd Molecular Weight (g/mol): 157.25 MDL Number: MFCD00011022 InChI Key: UIWYJDYFSGRHKR-UHFFFAOYSA-N Synonym: atom,gadolinio,unii-au0v1lm3jt,trihydride,au0v1lm3jt,foil, 3n,gadolinium, elemental,ingot,gadolinium, chips,compounds PubChem CID: 23982 ChEBI: CHEBI:33375 IUPAC Name: gadolinium SMILES: [Gd]
Gadolinium foil. 0.075mm (0.003in) thick, REacton™, 99.9% (REO)
CAS: 7440-54-2 Molecular Formula: Gd Molecular Weight (g/mol): 157.25 MDL Number: MFCD00011022 InChI Key: UIWYJDYFSGRHKR-UHFFFAOYSA-N Synonym: atom,gadolinio,unii-au0v1lm3jt,trihydride,au0v1lm3jt,foil, 3n,gadolinium, elemental,ingot,gadolinium, chips,compounds PubChem CID: 23982 ChEBI: CHEBI:33375 IUPAC Name: gadolinium SMILES: [Gd]
Thermo Scientific™ Gadolinium ingot, 99.9% (REM)
CAS: 7440-54-2 Molecular Formula: Gd Molecular Weight (g/mol): 157.25 MDL Number: MFCD00011022 InChI Key: UIWYJDYFSGRHKR-UHFFFAOYSA-N IUPAC Name: gadolinium SMILES: [Gd]
Gadolinium foil, 0.62mm (0.024in) thick, 99.9% (REO)
CAS: 7440-54-2 Molecular Formula: Gd Molecular Weight (g/mol): 157.25 MDL Number: MFCD00011022 InChI Key: UIWYJDYFSGRHKR-UHFFFAOYSA-N Synonym: atom,gadolinio,unii-au0v1lm3jt,trihydride,au0v1lm3jt,foil, 3n,gadolinium, elemental,ingot,gadolinium, chips,compounds PubChem CID: 23982 ChEBI: CHEBI:33375 IUPAC Name: gadolinium SMILES: [Gd]
Gadolinium foil, 0.025mm (0.001in) thick, 99.9% (REO)
CAS: 7440-54-2 Molecular Formula: Gd Molecular Weight (g/mol): 157.25 MDL Number: MFCD00011022 InChI Key: UIWYJDYFSGRHKR-UHFFFAOYSA-N Synonym: atom,gadolinio,unii-au0v1lm3jt,trihydride,au0v1lm3jt,foil, 3n,gadolinium, elemental,ingot,gadolinium, chips,compounds PubChem CID: 23982 ChEBI: CHEBI:33375 IUPAC Name: gadolinium SMILES: [Gd]
Gadolinium sputtering target, 76.2mm dia x 3.18mm thick
CAS: 7440-54-2 Molecular Formula: Gd Molecular Weight (g/mol): 157.25 MDL Number: MFCD00011022 InChI Key: UIWYJDYFSGRHKR-UHFFFAOYSA-N Synonym: atom,gadolinio,unii-au0v1lm3jt,trihydride,au0v1lm3jt,foil, 3n,gadolinium, elemental,ingot,gadolinium, chips,compounds PubChem CID: 23982 ChEBI: CHEBI:33375 IUPAC Name: gadolinium SMILES: [Gd]