CAS RN 7440-66-6
Zinc, granular, 20 mesh
CAS: 7440-66-6 Molecular Formula: Zn Molecular Weight (g/mol): 65.38 MDL Number: MFCD00011291 InChI Key: HCHKCACWOHOZIP-UHFFFAOYSA-N Synonym: dust,cinc,powder,merrillite,rheinzink,jasad,blue powder,granular,zinc, elemental,emanay dust PubChem CID: 23994 ChEBI: CHEBI:30185 IUPAC Name: zinc SMILES: [Zn]
Zinc powder, -100 mesh, 99.9% (metals basis), Thermo Scientific Chemicals
CAS: 7440-66-6 Molecular Formula: Zn Molecular Weight (g/mol): 65.38 MDL Number: MFCD00011291 InChI Key: HCHKCACWOHOZIP-UHFFFAOYSA-N Synonym: dust,cinc,powder,merrillite,rheinzink,jasad,blue powder,granular,zinc, elemental,emanay dust PubChem CID: 23994 ChEBI: CHEBI:30185 IUPAC Name: zinc SMILES: [Zn]
Zinc powder, -100 mesh, 97+%, Thermo Scientific Chemicals
CAS: 7440-66-6 Molecular Formula: Zn Molecular Weight (g/mol): 65.38 MDL Number: MFCD00011291 InChI Key: HCHKCACWOHOZIP-UHFFFAOYSA-N Synonym: dust,cinc,powder,merrillite,rheinzink,jasad,blue powder,granular,zinc, elemental,emanay dust PubChem CID: 23994 ChEBI: CHEBI:30185 IUPAC Name: zinc SMILES: [Zn]
Zinc granules, ACS, -20 mesh, 99.8% min
CAS: 7440-66-6 Molecular Formula: Zn Molecular Weight (g/mol): 65.38 MDL Number: MFCD00011291 InChI Key: HCHKCACWOHOZIP-UHFFFAOYSA-N Synonym: dust,cinc,powder,merrillite,rheinzink,jasad,blue powder,granular,zinc, elemental,emanay dust PubChem CID: 23994 ChEBI: CHEBI:30185 IUPAC Name: zinc SMILES: [Zn]
Zinc mossy, 2.5cm (0.98in) & down, 99% (metals basis)
CAS: 7440-66-6 Molecular Formula: Zn Molecular Weight (g/mol): 65.38 MDL Number: MFCD00011291 InChI Key: HCHKCACWOHOZIP-UHFFFAOYSA-N Synonym: dust,cinc,powder,merrillite,rheinzink,jasad,blue powder,granular,zinc, elemental,emanay dust PubChem CID: 23994 ChEBI: CHEBI:30185 IUPAC Name: zinc SMILES: [Zn]
Zinc shot, 1-5mm (0.04-0.2in), 99.999% (metals basis)
CAS: 7440-66-6 Molecular Formula: Zn Molecular Weight (g/mol): 65.38 MDL Number: MFCD00011291 InChI Key: HCHKCACWOHOZIP-UHFFFAOYSA-N Synonym: dust,cinc,powder,merrillite,rheinzink,jasad,blue powder,granular,zinc, elemental,emanay dust PubChem CID: 23994 ChEBI: CHEBI:30185 IUPAC Name: zinc SMILES: [Zn]
Zinc foil, 0.25mm (0.01in) thick, 30cm (12in) wide, 99.98% (metals basis)
CAS: 7440-66-6 Molecular Formula: Zn Molecular Weight (g/mol): 65.38 MDL Number: MFCD00011291 InChI Key: HCHKCACWOHOZIP-UHFFFAOYSA-N Synonym: dust,cinc,powder,merrillite,rheinzink,jasad,blue powder,granular,zinc, elemental,emanay dust PubChem CID: 23994 ChEBI: CHEBI:30185 IUPAC Name: zinc SMILES: [Zn]
Zinc powder, average 5-8 micron, 97.5% (metals basis), Thermo Scientific Chemicals
CAS: 7440-66-6 Molecular Formula: Zn Molecular Weight (g/mol): 65.38 MDL Number: MFCD00011291 InChI Key: HCHKCACWOHOZIP-UHFFFAOYSA-N Synonym: dust,cinc,powder,merrillite,rheinzink,jasad,blue powder,granular,zinc, elemental,emanay dust PubChem CID: 23994 ChEBI: CHEBI:30185 IUPAC Name: zinc SMILES: [Zn]
Zinc, Granular, 20 Mesh, ACS Reagent Grade, Ricca Chemical
CAS: 7440-66-6 Molecular Formula: Zn Molecular Weight (g/mol): 65.38 MDL Number: MFCD00011291 InChI Key: HCHKCACWOHOZIP-UHFFFAOYSA-N Synonym: dust,cinc,powder,merrillite,rheinzink,jasad,blue powder,granular,zinc, elemental,emanay dust PubChem CID: 23994 ChEBI: CHEBI:30185 IUPAC Name: zinc SMILES: [Zn]
Zinc wire, 2.0mm (0.08in) dia, 99.95% (metals basis)
CAS: 7440-66-6 Molecular Formula: Zn Molecular Weight (g/mol): 65.38 MDL Number: MFCD00011291 InChI Key: HCHKCACWOHOZIP-UHFFFAOYSA-N Synonym: dust,cinc,powder,merrillite,rheinzink,jasad,blue powder,granular,zinc, elemental,emanay dust PubChem CID: 23994 ChEBI: CHEBI:30185 IUPAC Name: zinc SMILES: [Zn]
Zinc wire, 1.0mm (0.04in) dia, 99.9997% (metals basis)
CAS: 7440-66-6 Molecular Formula: Zn Molecular Weight (g/mol): 65.38 MDL Number: MFCD00011291 InChI Key: HCHKCACWOHOZIP-UHFFFAOYSA-N Synonym: dust,cinc,powder,merrillite,rheinzink,jasad,blue powder,granular,zinc, elemental,emanay dust PubChem CID: 23994 ChEBI: CHEBI:30185 IUPAC Name: zinc SMILES: [Zn]
Zinc foil, 0.25mm (0.01in) thick, Puratronic™, 99.994% (metals basis)
CAS: 7440-66-6 Molecular Formula: Zn Molecular Weight (g/mol): 65.38 MDL Number: MFCD00011291 InChI Key: HCHKCACWOHOZIP-UHFFFAOYSA-N Synonym: dust,cinc,powder,merrillite,rheinzink,jasad,blue powder,granular,zinc, elemental,emanay dust PubChem CID: 23994 ChEBI: CHEBI:30185 IUPAC Name: zinc SMILES: [Zn]
Zinc shot, 1-6mm (0.04-0.24in), Puratronic™, 99.9999% (metals basis)
CAS: 7440-66-6 Molecular Formula: Zn Molecular Weight (g/mol): 65.38 MDL Number: MFCD00011291 InChI Key: HCHKCACWOHOZIP-UHFFFAOYSA-N Synonym: dust,cinc,powder,merrillite,rheinzink,jasad,blue powder,granular,zinc, elemental,emanay dust PubChem CID: 23994 ChEBI: CHEBI:30185 IUPAC Name: zinc SMILES: [Zn]
Zinc Standard, 1000mg/L in 2% HNO3 (Assurance Grade), SPEX CertiPrep™
CAS: 7440-66-6 Molecular Formula: Zn Molecular Weight (g/mol): 65.38 MDL Number: MFCD00011291 InChI Key: HCHKCACWOHOZIP-UHFFFAOYSA-N PubChem CID: 23994 ChEBI: CHEBI:30185 IUPAC Name: zinc SMILES: [Zn]
Zinc, 10 w/v% suspension in THF, Thermo Scientific Chemicals
CAS: 7440-66-6 Molecular Formula: Zn Molecular Weight (g/mol): 65.38 MDL Number: MFCD00011291 InChI Key: HCHKCACWOHOZIP-UHFFFAOYSA-N IUPAC Name: zinc SMILES: [Zn]