accessibility menu, dialog, popup

UserName

Viewing profile as user:
Ca

CAS RN 7440-70-2

IUPAC Name: calcium
Synonyms: calcium|calcio|calcium(0)|kalzium|calcium atom|Can
Molecular Weight (g/mol): 40.08
Molecular Formula: Ca
InChi Key: OYPRJOBELJOOCE-UHFFFAOYSA-N
SMILES: [Ca]
Quantity 
  • (2)
  • (2)
  • (5)
  • (6)
  • (3)
  • (5)
  • (2)
  • (4)
  • (1)
Form 
  • (10)
  • (1)
  • (2)
  • (5)
Percent Purity 
  • (2)
  • (6)
  • (1)
  • (1)
  • (2)
  • (2)
Grade 
  • (2)
  • (8)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (8)
  • (12)
  • (20)
  • (4)
  • (5)
  • (3)
  • (2)
  • (2)
  • (5)
  • (6)
  • (3)
  • (5)
  • (2)
  • (4)
  • (1)
  • (10)
  • (1)
  • (2)
  • (5)
  • (2)
  • (6)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (8)
  • (1)
  • (2)
  • (1)
  • (4)
  • (1)
  • (3)
  • (2)
  • (1)
  • (8)
  • (8)
  • (1)
  • (1)
  • (4)
  • (2)
  • (2)