accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 74409-52-2

H3CNNONONCH3Cl

CAS RN 74409-52-2

IUPAC Name: 1-(3-chloropropyl)-3,7-dimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione
Synonyms: 1-(3-chloropropyl)-3,7-dimethylpurine-2,6-dione
Molecular Weight (g/mol): 256.69
Molecular Formula: C10H13ClN4O2
InChi Key: JMOWKXFNJSTXBW-UHFFFAOYSA-N
SMILES: CN1C=NC2=C1C(=O)N(CCCCl)C(=O)N2C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results