accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 74431-23-5

IUPAC Name: (5R,6S)-3-({2-[(aminomethylidene)amino]ethyl}sulfanyl)-6-[(1S)-1-hydroxyethyl]-7-oxo-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylic acid hydrate
Synonyms: imipenem
Molecular Weight (g/mol): 317.36
Molecular Formula: C12H19N3O5S
InChi Key: GSOSVVULSKVSLQ-XMCAPODCSA-N
SMILES: O.C[C@H](O)[C@@H]1[C@H]2CC(SCCN=CN)=C(N2C1=O)C(O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Type 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (2)
  • (1)