CAS RN 7446-08-4
Selenium(IV) oxide, 99.4% (metals basis)
CAS: 7446-08-4 Molecular Formula: O2Se Molecular Weight (g/mol): 110.97 MDL Number: MFCD00003562 InChI Key: JPJALAQPGMAKDF-UHFFFAOYSA-N Synonym: selenium oxide,selenium iv oxide,oxoselane oxide,selenious anhydride,selenium oxide seo2,seleniumdioxide,dimer,selenous acid anhydride,selenium iv dioxide 1:2,selenium oxide se2o4 PubChem CID: 24007 SMILES: O=[Se]=O
Selenium(IV) oxide, Puratronic™, 99.999% (metals basis)
CAS: 7446-08-4 Molecular Formula: O2Se Molecular Weight (g/mol): 110.97 MDL Number: MFCD00003562 InChI Key: JPJALAQPGMAKDF-UHFFFAOYSA-N Synonym: selenium oxide,selenium iv oxide,oxoselane oxide,selenious anhydride,selenium oxide seo2,seleniumdioxide,dimer,selenous acid anhydride,selenium iv dioxide 1:2,selenium oxide se2o4 PubChem CID: 24007 IUPAC Name: selenium dioxide SMILES: O=[Se]=O
Selenium(IV) oxide, 99.999%, (trace metal basis)
CAS: 7446-08-4 Molecular Formula: O2Se Molecular Weight (g/mol): 110.97 MDL Number: MFCD00003562 InChI Key: JPJALAQPGMAKDF-UHFFFAOYSA-N Synonym: selenium oxide,selenium iv oxide,oxoselane oxide,selenious anhydride,selenium oxide seo2,seleniumdioxide,dimer,selenous acid anhydride,selenium iv dioxide 1:2,selenium oxide se2o4 PubChem CID: 24007 IUPAC Name: selenium dioxide SMILES: O=[Se]=O
Selenium(IV) oxide, 99.8%, (trace metal basis)
CAS: 7446-08-4 Molecular Formula: O2Se Molecular Weight (g/mol): 110.97 MDL Number: MFCD00003562 InChI Key: JPJALAQPGMAKDF-UHFFFAOYSA-N Synonym: selenium oxide,selenium iv oxide,oxoselane oxide,selenious anhydride,selenium oxide seo2,seleniumdioxide,dimer,selenous acid anhydride,selenium iv dioxide 1:2,selenium oxide se2o4 PubChem CID: 24007 SMILES: O=[Se]=O
Selenium AA Standard, 1000 ppm Se in H2O, Ricca Chemical
CAS: 7446-08-4 Molecular Formula: O2Se Molecular Weight (g/mol): 110.97 MDL Number: MFCD00003562 InChI Key: JPJALAQPGMAKDF-UHFFFAOYSA-N PubChem CID: 24007 IUPAC Name: (oxo-λ4-selanylidene)oxidane SMILES: O=[Se]=O
Selenium Standard For ICP/MS, SPEX CertiPrep™
CAS: 7446-08-4 Molecular Formula: O2Se Molecular Weight (g/mol): 110.97 MDL Number: MFCD00003562 InChI Key: JPJALAQPGMAKDF-UHFFFAOYSA-N PubChem CID: 24007 IUPAC Name: selenium dioxide SMILES: O=[Se]=O
Sigma Aldrich Selenium Dioxide
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Selenium Dioxide
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Selenium(IV) Oxide p.a., Thermo Scientific™
CAS: 7446-08-4 Molecular Formula: O2Se Molecular Weight (g/mol): 110.97 MDL Number: MFCD00003562 InChI Key: JPJALAQPGMAKDF-UHFFFAOYSA-N Synonym: selenium oxide,selenium iv oxide,oxoselane oxide,selenious anhydride,selenium oxide seo2,seleniumdioxide,dimer,selenous acid anhydride,selenium iv dioxide 1:2,selenium oxide se2o4 PubChem CID: 24007 IUPAC Name: (oxo-λ⁴-selanylidene)oxidane SMILES: O=[Se]=O
Ricca Chemical Company Selenium Standard, 1mL = 1mg Se, 1000ppm Se, Ricca Chemical
CAS: 7446-08-4 Molecular Formula: O2Se Molecular Weight (g/mol): 110.97 MDL Number: MFCD00003562 InChI Key: JPJALAQPGMAKDF-UHFFFAOYSA-N PubChem CID: 24007 IUPAC Name: (oxo-λ⁴-selanylidene)oxidane SMILES: O=[Se]=O