accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7446-33-5

IUPAC Name: diyttrium(3+) octahydrate trisulfate
Synonyms: diyttrium(3+) octahydrate trisulfate
Molecular Weight (g/mol): 610.1
Molecular Formula: H16O20S3Y2
InChi Key: OMBXBIHFXZAMLA-UHFFFAOYSA-H
SMILES: O.O.O.O.O.O.O.O.[Y+3].[Y+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Grade 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results