CAS RN 7446-70-0
Aluminum chloride, anhydrous, granular, 99%
CAS: 7446-70-0 Molecular Formula: AlCl3 Molecular Weight (g/mol): 133.33 MDL Number: MFCD00003422 InChI Key: VSCWAEJMTAWNJL-UHFFFAOYSA-K Synonym: aluminum chloride anhydrous,aluminum iii chloride,aluminiumchloride,aluminumchloride,aluminum chlorid,aluminum cloride,alumimum chloride,aluminum-chloride,trichloroaluminium,chloride aluminum IUPAC Name: aluminum(3+) trichloride SMILES: [Al+3].[Cl-].[Cl-].[Cl-]
Aluminum chloride, 99%, extra pure, anhydrous, granules
CAS: 7446-70-0 Molecular Formula: AlCl3 Molecular Weight (g/mol): 133.33 MDL Number: MFCD00003422 InChI Key: VSCWAEJMTAWNJL-UHFFFAOYSA-K Synonym: aluminum chloride anhydrous,aluminum iii chloride,aluminiumchloride,aluminumchloride,aluminum chlorid,aluminum cloride,alumimum chloride,aluminum-chloride,trichloroaluminium,chloride aluminum IUPAC Name: aluminum(3+) trichloride SMILES: [Al+3].[Cl-].[Cl-].[Cl-]
Aluminum chloride, 6.0N Standardized Solution
CAS: 7446-70-0 Molecular Formula: AlCl3 Molecular Weight (g/mol): 133.33 MDL Number: MFCD00003422 InChI Key: VSCWAEJMTAWNJL-UHFFFAOYSA-K IUPAC Name: aluminum(3+) trichloride SMILES: [Al+3].[Cl-].[Cl-].[Cl-]
Aluminum chloride, 98.5%, extra pure, anhydrous, powder
CAS: 7446-70-0 Molecular Formula: AlCl3 Molecular Weight (g/mol): 133.33 MDL Number: MFCD00003422 InChI Key: VSCWAEJMTAWNJL-UHFFFAOYSA-K Synonym: aluminum chloride anhydrous,aluminum iii chloride,aluminiumchloride,aluminumchloride,aluminum chlorid,aluminum cloride,alumimum chloride,aluminum-chloride,trichloroaluminium,chloride aluminum IUPAC Name: aluminum(3+) trichloride SMILES: [Al+3].[Cl-].[Cl-].[Cl-]
Aluminum chloride, 99.999%, (trace metal basis), for analysis, anhydrous, powder
CAS: 7446-70-0 Molecular Formula: AlCl3 Molecular Weight (g/mol): 133.33 MDL Number: MFCD00003422 InChI Key: VSCWAEJMTAWNJL-UHFFFAOYSA-K Synonym: aluminum chloride anhydrous,aluminum iii chloride,aluminiumchloride,aluminumchloride,aluminum chlorid,aluminum cloride,alumimum chloride,aluminum-chloride,trichloroaluminium,chloride aluminum IUPAC Name: aluminum(3+) trichloride SMILES: [Al+3].[Cl-].[Cl-].[Cl-]
Aluminum chloride, ultra dry, 99.999% (metals basis)
CAS: 7446-70-0 Molecular Formula: AlCl3 Molecular Weight (g/mol): 133.33 MDL Number: MFCD00003422 InChI Key: VSCWAEJMTAWNJL-UHFFFAOYSA-K Synonym: aluminum chloride anhydrous,aluminum iii chloride,aluminiumchloride,aluminumchloride,aluminum chlorid,aluminum cloride,alumimum chloride,aluminum-chloride,trichloroaluminium,chloride aluminum IUPAC Name: aluminum(3+) trichloride SMILES: [Al+3].[Cl-].[Cl-].[Cl-]
Aluminum chloride, 98+%, extra pure, anhydrous powder
CAS: 7446-70-0 Molecular Formula: AlCl3 Molecular Weight (g/mol): 133.33 MDL Number: MFCD00003422 InChI Key: VSCWAEJMTAWNJL-UHFFFAOYSA-K Synonym: aluminum chloride anhydrous,aluminum iii chloride,aluminiumchloride,aluminumchloride,aluminum chlorid,aluminum cloride,alumimum chloride,aluminum-chloride,trichloroaluminium,chloride aluminum IUPAC Name: aluminum(3+) trichloride SMILES: [Al+3].[Cl-].[Cl-].[Cl-]
Aluminum chloride, anhydrous, 99.985% (metals basis)
CAS: 7446-70-0 Molecular Formula: AlCl3 Molecular Weight (g/mol): 133.33 MDL Number: MFCD00003422 InChI Key: VSCWAEJMTAWNJL-UHFFFAOYSA-K Synonym: aluminum chloride anhydrous,aluminum iii chloride,aluminiumchloride,aluminumchloride,aluminum chlorid,aluminum cloride,alumimum chloride,aluminum-chloride,trichloroaluminium,chloride aluminum IUPAC Name: aluminum(3+) trichloride SMILES: [Al+3].[Cl-].[Cl-].[Cl-]
Aluminum chloride, anhydrous, Reagent Grade
CAS: 7446-70-0 Molecular Formula: AlCl3 Molecular Weight (g/mol): 133.33 MDL Number: MFCD00003422 InChI Key: VSCWAEJMTAWNJL-UHFFFAOYSA-K Synonym: aluminum chloride anhydrous,aluminum iii chloride,aluminiumchloride,aluminumchloride,aluminum chlorid,aluminum cloride,alumimum chloride,aluminum-chloride,trichloroaluminium,chloride aluminum IUPAC Name: aluminum(3+) trichloride SMILES: [Al+3].[Cl-].[Cl-].[Cl-]
Aluminum chloride, anhydrous, 99.999% (metals basis)
CAS: 7446-70-0 Molecular Formula: AlCl3 Molecular Weight (g/mol): 133.33 MDL Number: MFCD00003422 InChI Key: VSCWAEJMTAWNJL-UHFFFAOYSA-K Synonym: aluminum chloride anhydrous,aluminum iii chloride,aluminiumchloride,aluminumchloride,aluminum chlorid,aluminum cloride,alumimum chloride,aluminum-chloride,trichloroaluminium,chloride aluminum IUPAC Name: aluminum(3+) trichloride SMILES: [Al+3].[Cl-].[Cl-].[Cl-]
Aluminum chloride, ultra dry, 99.99% (metals basis)
CAS: 7446-70-0 Molecular Formula: AlCl3 Molecular Weight (g/mol): 133.33 MDL Number: MFCD00003422 InChI Key: VSCWAEJMTAWNJL-UHFFFAOYSA-K Synonym: aluminum chloride anhydrous,aluminum iii chloride,aluminiumchloride,aluminumchloride,aluminum chlorid,aluminum cloride,alumimum chloride,aluminum-chloride,trichloroaluminium,chloride aluminum IUPAC Name: aluminum(3+) trichloride SMILES: [Al+3].[Cl-].[Cl-].[Cl-]
Aluminum chloride, anhydrous, 95+%
CAS: 7446-70-0 Molecular Formula: AlCl3 Molecular Weight (g/mol): 133.33 MDL Number: MFCD00003422 InChI Key: VSCWAEJMTAWNJL-UHFFFAOYSA-K Synonym: aluminum chloride anhydrous,aluminum iii chloride,aluminiumchloride,aluminumchloride,aluminum chlorid,aluminum cloride,alumimum chloride,aluminum-chloride,trichloroaluminium,chloride aluminum IUPAC Name: aluminum(3+) trichloride SMILES: [Al+3].[Cl-].[Cl-].[Cl-]
Aluminum(III) Chloride 98.0+%, TCI America™
CAS: 7446-70-0 Molecular Formula: AlCl3 Molecular Weight (g/mol): 133.33 MDL Number: MFCD00003422 InChI Key: VSCWAEJMTAWNJL-UHFFFAOYSA-K Synonym: aluminum chloride anhydrous,aluminum iii chloride,aluminiumchloride,aluminumchloride,aluminum chlorid,aluminum cloride,alumimum chloride,aluminum-chloride,trichloroaluminium,chloride aluminum IUPAC Name: aluminum(3+) trichloride SMILES: [Al+3].[Cl-].[Cl-].[Cl-]
VeriSpec™ Aluminum Standard for ICP 1000 ppm in 2% HCl, Ricca Chemical
CAS: 7446-70-0 Molecular Formula: AlCl3 Molecular Weight (g/mol): 133.33 MDL Number: MFCD00003422 InChI Key: VSCWAEJMTAWNJL-UHFFFAOYSA-K IUPAC Name: aluminum(3+) trichloride SMILES: [Al+3].[Cl-].[Cl-].[Cl-]
- 1
- 2