accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSNOO

CAS RN 74470-23-8

IUPAC Name: 2-(methylsulfanyl)pyridine-3-carboxylate
Synonyms: 2-(methylsulfanyl)pyridine-3-carboxylate
Molecular Weight (g/mol): 168.19
Molecular Formula: C7H6NO2S
InChi Key: COPSJQVPEUUOKY-UHFFFAOYSA-M
SMILES: CSC1=NC=CC=C1C([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)