accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3FFFCl

CAS RN 74483-48-0

IUPAC Name: 2-chloro-1-methyl-3-(trifluoromethyl)benzene
Synonyms: 2-chloro-1-methyl-3-(trifluoromethyl)benzene
Molecular Weight (g/mol): 194.58
Molecular Formula: C8H6ClF3
InChi Key: NJLAMXMEANIPBX-UHFFFAOYSA-N
SMILES: CC1=C(Cl)C(=CC=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results