accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 745038-86-2

IUPAC Name: lithium(1+) chloro(propan-2-yl)magnesium chloride
Synonyms: lithium(1+) isopropylmagnesium chloride chloride
Molecular Weight (g/mol): 145.23
Molecular Formula: C3H7Cl2LiMg
InChi Key: CWTUREABAILGIK-UHFFFAOYSA-L
SMILES: [Li+].[Cl-].CC(C)[Mg]Cl
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)