accessibility menu, dialog, popup

UserName

Viewing profile as user:
LiBHCH3H3CH3C

CAS RN 74540-86-6

IUPAC Name: lithium(1+) triethylboranuide
Synonyms: lithium triethylborohydride
Molecular Weight (g/mol): 105.94
Molecular Formula: C6H16BLi
InChi Key: WCJAYABJWDIZAJ-UHFFFAOYSA-N
SMILES: [Li+].CC[BH-](CC)CC
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results