accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOHHOOH

CAS RN 74568-07-3

IUPAC Name: pentacyclo[19.3.1.1³,⁷.1⁹,¹³.1¹⁵,¹⁹]octacosa-1(25),3,5,7(28),9,11,13(27),15,17,19(26),21,23-dodecaene-25,26,27,28-tetrol
Synonyms: pentacyclo[19.3.1.1³,⁷.1⁹,¹³.1¹⁵,¹⁹]octacosa-1(25),3,5,7(28),9,11,13(27),15,17,19(26),21,23-dodecaene-25,26,27,28-tetrol
Molecular Weight (g/mol): 424.5
Molecular Formula: C28H24O4
InChi Key: YPNHVQZZPXPQOS-UHFFFAOYSA-N
SMILES: OC1=C2CC3=CC=CC(CC4=CC=CC(CC5=CC=CC(CC1=CC=C2)=C5O)=C4O)=C3O
Molecular Weight (g/mol) 
  • (1)
  • (5)
  • (2)
Quantity 
  • (4)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (4)
  • (1)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (5)
  • (1)
  • (5)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (2)
  • (4)
  • (1)
  • (4)