accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 745783-87-3

IUPAC Name: 2-isocyanatooctane
Synonyms: 2-isocyanatooctane
Molecular Weight (g/mol): 155.24
Molecular Formula: C9H17NO
InChi Key: FTWPDIQXUDUDPL-UHFFFAOYNA-N
SMILES: CCCCCCC(C)N=C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results