accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOFFF

CAS RN 745783-91-9

IUPAC Name: 2-oxo-2-[2-(trifluoromethyl)phenyl]acetaldehyde
Synonyms: 2-oxo-2-[2-(trifluoromethyl)phenyl]acetaldehyde
Molecular Weight (g/mol): 202.13
Molecular Formula: C9H5F3O2
InChi Key: HBOSPPHIPGYHDI-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=CC=C1C(=O)C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results