accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH2Br

CAS RN 74586-53-1

IUPAC Name: 3-bromo-5-methylaniline
Synonyms: 3-bromo-5-methylaniline
Molecular Weight (g/mol): 186.05
Molecular Formula: C7H8BrN
InChi Key: YIZRPAWCIFTHNA-UHFFFAOYSA-N
SMILES: CC1=CC(N)=CC(Br)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results