accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 74587-12-5

IUPAC Name: 3-iodo-4-methoxyaniline
Synonyms: 3-iodo-4-methoxyaniline
Molecular Weight (g/mol): 249.05
Molecular Formula: C7H8INO
InChi Key: UHPNLGCUIGEZRB-UHFFFAOYSA-N
SMILES: COC1=C(I)C=C(N)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results