CAS RN 7462-73-9
IUPAC Name:
2-chlorobutanamide
Synonyms:
2-chlorobutanamide
Molecular Weight (g/mol):
121.56
Molecular Formula:
C4H8ClNO
InChi Key:
VJIOSCMTZVXQQU-UHFFFAOYNA-N
SMILES:
CCC(Cl)C(N)=O
1
–
1
of
1
results
1
Promotions Available
2-Chlorobutyramide, 98%, Thermo Scientific Chemicals
2-Chlorobutyramide, 98%, Thermo Scientific Chemicals
CAS: 7462-73-9 Molecular Formula: C4H8ClNO Molecular Weight (g/mol): 121.564 MDL Number: MFCD00041477 InChI Key: VJIOSCMTZVXQQU-UHFFFAOYSA-N Synonym: 2-chlorobutyramide PubChem CID: 344969 IUPAC Name: 2-chlorobutanamide SMILES: CCC(C(=O)N)Cl