accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7465-88-5

IUPAC Name: 4-methoxy-N-phenylbenzamide
Synonyms: 4-methoxy-N-phenylbenzamide
Molecular Weight (g/mol): 227.26
Molecular Formula: C14H13NO2
InChi Key: XOJAJRFBOKCXPI-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C(=O)NC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results