accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7466-54-8

IUPAC Name: 2-methoxybenzohydrazide
Synonyms: benzoylhydrazine, O-methoxy
Molecular Weight (g/mol): 166.18
Molecular Formula: C8H10N2O2
InChi Key: QMGXWNSSMGAHCA-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1C(=O)NN
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results